Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49432574-49432835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC17A7
Entrez Gene ID
57030
Gene Name
solute carrier family 17 member 7
Gene Aliases
BNPI, VGLUT1
UniGene
Hs.375616
Gene Chromosome Location
Chr.19: 49429398-49441551 on Build GRCh38
Transcripts
NM_020309.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
999292 c.1091T>G p.F364C Chr.19: 49432578-49432578 on Build GRCh38
3835375 c.1055C>T p.T352I Chr.19: 49432614-49432614 on Build GRCh38
6266005 c.1068C>A p.P356P Chr.19: 49432601-49432601 on Build GRCh38
267431 c.999C>T p.F333F Chr.19: 49432829-49432829 on Build GRCh38
4726597 c.1084G>C p.A362P Chr.19: 49432585-49432585 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759357519 Chr.19: 49432800-49432800 on Build GRCh38
rs5828394 Chr.19: 49432671-49432672 on Build GRCh38
rs776405254 Chr.19: 49432655-49432655 on Build GRCh38
rs757398396 Chr.19: 49432692-49432692 on Build GRCh38
rs1007777906 Chr.19: 49432636-49432636 on Build GRCh38
rs202161365 Chr.19: 49432657-49432657 on Build GRCh38
rs548390425 Chr.19: 49432720-49432720 on Build GRCh38
rs766348339 Chr.19: 49432684-49432684 on Build GRCh38
rs1004920861 Chr.19: 49432752-49432752 on Build GRCh38
rs772883984 Chr.19: 49432784-49432784 on Build GRCh38
rs1018886200 Chr.19: 49432654-49432654 on Build GRCh38
rs59192444 Chr.19: 49432733-49432733 on Build GRCh38
rs529942781 Chr.19: 49432718-49432718 on Build GRCh38
rs764845971 Chr.19: 49432665-49432665 on Build GRCh38
rs200536682 Chr.19: 49432791-49432791 on Build GRCh38
rs112616652 Chr.19: 49432687-49432687 on Build GRCh38
rs772572486 Chr.19: 49432697-49432697 on Build GRCh38
rs776832877 Chr.19: 49432765-49432765 on Build GRCh38
rs761578221 Chr.19: 49432767-49432767 on Build GRCh38
rs774341076 Chr.19: 49432801-49432802 on Build GRCh38
rs769133255 Chr.19: 49432703-49432703 on Build GRCh38
rs776779258 Chr.19: 49432698-49432698 on Build GRCh38
rs761370168 Chr.19: 49432664-49432664 on Build GRCh38
rs748266202 Chr.19: 49432789-49432789 on Build GRCh38
rs372417324 Chr.19: 49432643-49432643 on Build GRCh38
rs568440103 Chr.19: 49432670-49432671 on Build GRCh38
rs564881392 Chr.19: 49432598-49432598 on Build GRCh38
rs59848838 Chr.19: 49432779-49432779 on Build GRCh38
rs112546257 Chr.19: 49432706-49432706 on Build GRCh38
rs892135680 Chr.19: 49432768-49432768 on Build GRCh38
rs1016270411 Chr.19: 49432755-49432755 on Build GRCh38
rs768928158 Chr.19: 49432629-49432629 on Build GRCh38
rs398034942 Chr.19: 49432673-49432674 on Build GRCh38
rs754127454 Chr.19: 49432691-49432691 on Build GRCh38
rs778985851 Chr.19: 49432693-49432693 on Build GRCh38
rs532411812 Chr.19: 49432619-49432619 on Build GRCh38
rs745908258 Chr.19: 49432694-49432694 on Build GRCh38
rs763226791 Chr.19: 49432672-49432673 on Build GRCh38
rs202191497 Chr.19: 49432700-49432700 on Build GRCh38
rs749935315 Chr.19: 49432680-49432680 on Build GRCh38
rs1014479172 Chr.19: 49432719-49432719 on Build GRCh38
rs367721849 Chr.19: 49432686-49432686 on Build GRCh38
rs764286365 Chr.19: 49432782-49432782 on Build GRCh38
rs148186322 Chr.19: 49432595-49432595 on Build GRCh38
rs374987041 Chr.19: 49432786-49432786 on Build GRCh38
rs375290856 Chr.19: 49432685-49432685 on Build GRCh38
rs928278024 Chr.19: 49432721-49432721 on Build GRCh38

View Full Product Details