Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 107746610-107746883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCML4
Entrez Gene ID
256380
Gene Name
sex comb on midleg-like 4 (Drosophila)
Gene Aliases
dJ47M23.1
UniGene
Hs.486109
Gene Chromosome Location
Chr.6: 107697297-107845959 on Build GRCh38
Transcripts
NM_001286408.1 NM_198081.4 XM_011535703.2 XM_011535704.2 XM_011535706.1 XM_011535707.2 XM_011535710.2 XM_017010677.1 XM_017010678.1 XM_017010679.1 XM_017010680.1 XM_017010681.1 XM_017010682.1 XM_017010683.1 XM_017010684.1 XM_017010685.1 XM_017010686.1 XM_017010687.1 XM_017010688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4487957 c.237C>T p.P79P Chr.6: 107746852-107746852 on Build GRCh38
6772784 c.400+2T>C p.? Chr.6: 107746687-107746687 on Build GRCh38
4416710 c.290G>A p.R97Q Chr.6: 107746799-107746799 on Build GRCh38
2150706 c.302delT p.V101fs*50 Chr.6: 107746787-107746787 on Build GRCh38
6772774 c.231G>A p.A77A Chr.6: 107746858-107746858 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs6934505 Chr.6: 107746799-107746799 on Build GRCh38
rs759394869 Chr.6: 107746854-107746854 on Build GRCh38
rs909086109 Chr.6: 107746814-107746814 on Build GRCh38
rs746008202 Chr.6: 107746727-107746727 on Build GRCh38
rs975818673 Chr.6: 107746789-107746789 on Build GRCh38
rs780363936 Chr.6: 107746800-107746800 on Build GRCh38
rs201979284 Chr.6: 107746788-107746788 on Build GRCh38
rs143639662 Chr.6: 107746860-107746860 on Build GRCh38
rs772784470 Chr.6: 107746752-107746752 on Build GRCh38
rs940697403 Chr.6: 107746846-107746846 on Build GRCh38
rs879934792 Chr.6: 107746700-107746700 on Build GRCh38
rs747405400 Chr.6: 107746733-107746733 on Build GRCh38
rs772795692 Chr.6: 107746646-107746646 on Build GRCh38
rs200161335 Chr.6: 107746764-107746764 on Build GRCh38
rs748589627 Chr.6: 107746816-107746816 on Build GRCh38
rs1005282693 Chr.6: 107746678-107746678 on Build GRCh38
rs41287522 Chr.6: 107746711-107746711 on Build GRCh38
rs201217173 Chr.6: 107746724-107746724 on Build GRCh38
rs775436252 Chr.6: 107746859-107746859 on Build GRCh38
rs757463631 Chr.6: 107746722-107746722 on Build GRCh38
rs776790300 Chr.6: 107746743-107746743 on Build GRCh38
rs751899100 Chr.6: 107746804-107746804 on Build GRCh38
rs766078524 Chr.6: 107746758-107746758 on Build GRCh38
rs751384036 Chr.6: 107746674-107746674 on Build GRCh38
rs370055138 Chr.6: 107746767-107746767 on Build GRCh38
rs748864966 Chr.6: 107746830-107746830 on Build GRCh38
rs762804924 Chr.6: 107746658-107746658 on Build GRCh38
rs375948039 Chr.6: 107746753-107746753 on Build GRCh38
rs758655825 Chr.6: 107746729-107746729 on Build GRCh38
rs760664220 Chr.6: 107746787-107746787 on Build GRCh38
rs767543814 Chr.6: 107746784-107746784 on Build GRCh38
rs769256462 Chr.6: 107746750-107746750 on Build GRCh38
rs781661002 Chr.6: 107746813-107746813 on Build GRCh38
rs1013724845 Chr.6: 107746697-107746697 on Build GRCh38
rs769120247 Chr.6: 107746741-107746741 on Build GRCh38
rs372302614 Chr.6: 107746803-107746803 on Build GRCh38
rs774252439 Chr.6: 107746776-107746776 on Build GRCh38
rs755393210 Chr.6: 107746811-107746811 on Build GRCh38
rs896533376 Chr.6: 107746692-107746692 on Build GRCh38
rs555798944 Chr.6: 107746732-107746732 on Build GRCh38
rs372377654 Chr.6: 107746749-107746749 on Build GRCh38
rs774198851 Chr.6: 107746841-107746841 on Build GRCh38
rs142985964 Chr.6: 107746703-107746703 on Build GRCh38
rs750535686 Chr.6: 107746798-107746798 on Build GRCh38
rs867871502 Chr.6: 107746629-107746629 on Build GRCh38
rs536132560 Chr.6: 107746713-107746713 on Build GRCh38
rs770125898 Chr.6: 107746668-107746668 on Build GRCh38
rs752742400 Chr.6: 107746786-107746786 on Build GRCh38
rs374332910 Chr.6: 107746660-107746660 on Build GRCh38
rs770462390 Chr.6: 107746834-107746834 on Build GRCh38
rs770348336 Chr.6: 107746820-107746820 on Build GRCh38
rs371827897 Chr.6: 107746858-107746858 on Build GRCh38
rs1014968445 Chr.6: 107746681-107746681 on Build GRCh38
rs777099113 Chr.6: 107746822-107746822 on Build GRCh38
rs201534795 Chr.6: 107746795-107746795 on Build GRCh38
rs759448219 Chr.6: 107746780-107746780 on Build GRCh38
rs887821615 Chr.6: 107746631-107746631 on Build GRCh38
rs1032998992 Chr.6: 107746718-107746718 on Build GRCh38

View Full Product Details