Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7627194-7627421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAT2
Entrez Gene ID
112483
Gene Name
spermidine/spermine N1-acetyltransferase family member 2
Gene Aliases
SSAT2
UniGene
Hs.10846
Gene Chromosome Location
Chr.17: 7626234-7627878 on Build GRCh38
Transcripts
NM_001320845.1 NM_001320846.1 NM_001320847.1 NM_133491.4 XM_017024073.1 XM_017024074.1 NM_001289114.1

Gene Symbol
SHBG
Entrez Gene ID
6462
Gene Name
sex hormone binding globulin
Gene Aliases
ABP, SBP, TEBG
UniGene
Hs.632235
Gene Chromosome Location
Chr.17: 7614064-7633383 on Build GRCh38
Transcripts
NM_001320845.1 NM_001320846.1 NM_001320847.1 NM_133491.4 XM_017024073.1 XM_017024074.1 NM_001289114.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5178828 c.118+7G>T p.? Chr.17: 7627356-7627356 on Build GRCh38
6571442 c.125G>A p.R42K Chr.17: 7627220-7627220 on Build GRCh38
3712575 c.123G>A p.L41L Chr.17: 7627222-7627222 on Build GRCh38
7349457 c.121delC p.L41fs*1 Chr.17: 7627224-7627224 on Build GRCh38
5780612 c.97C>T p.Q33* Chr.17: 7627384-7627384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748937151 Chr.17: 7627366-7627366 on Build GRCh38
rs757584878 Chr.17: 7627233-7627233 on Build GRCh38
rs758727744 Chr.17: 7627270-7627270 on Build GRCh38
rs770693140 Chr.17: 7627372-7627372 on Build GRCh38
rs780460227 Chr.17: 7627317-7627317 on Build GRCh38
rs1002375114 Chr.17: 7627313-7627313 on Build GRCh38
rs769275716 Chr.17: 7627361-7627361 on Build GRCh38
rs1013988186 Chr.17: 7627389-7627389 on Build GRCh38
rs376908982 Chr.17: 7627344-7627344 on Build GRCh38
rs370121688 Chr.17: 7627222-7627222 on Build GRCh38
rs900657041 Chr.17: 7627227-7627227 on Build GRCh38
rs767450293 Chr.17: 7627390-7627390 on Build GRCh38
rs774139909 Chr.17: 7627387-7627387 on Build GRCh38
rs896743827 Chr.17: 7627332-7627332 on Build GRCh38
rs1026335240 Chr.17: 7627352-7627352 on Build GRCh38
rs766160544 Chr.17: 7627249-7627250 on Build GRCh38
rs373867779 Chr.17: 7627325-7627325 on Build GRCh38
rs759398278 Chr.17: 7627388-7627388 on Build GRCh38
rs372422201 Chr.17: 7627340-7627340 on Build GRCh38
rs753902555 Chr.17: 7627225-7627225 on Build GRCh38
rs1049299972 Chr.17: 7627281-7627281 on Build GRCh38
rs750651025 Chr.17: 7627267-7627267 on Build GRCh38
rs930926323 Chr.17: 7627229-7627229 on Build GRCh38
rs772895399 Chr.17: 7627365-7627365 on Build GRCh38
rs947144063 Chr.17: 7627220-7627220 on Build GRCh38
rs751412162 Chr.17: 7627389-7627390 on Build GRCh38
rs747614313 Chr.17: 7627348-7627348 on Build GRCh38
rs377569695 Chr.17: 7627353-7627353 on Build GRCh38
rs765505793 Chr.17: 7627254-7627254 on Build GRCh38
rs755452318 Chr.17: 7627330-7627330 on Build GRCh38
rs11654494 Chr.17: 7627351-7627351 on Build GRCh38

View Full Product Details