Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 32096943-32097194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RCN1
Entrez Gene ID
5954
Gene Name
reticulocalbin 1
Gene Aliases
HEL-S-84, PIG20, RCAL, RCN
UniGene
Hs.97887
Gene Chromosome Location
Chr.11: 32090931-32105726 on Build GRCh38
Transcripts
NM_002901.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
926353 c.303T>C p.T101T Chr.11: 32097192-32097192 on Build GRCh38
4191057 c.260T>C p.I87T Chr.11: 32097149-32097149 on Build GRCh38
7421806 c.280G>A p.D94N Chr.11: 32097169-32097169 on Build GRCh38
926352 c.268C>T p.R90* Chr.11: 32097157-32097157 on Build GRCh38
284528 c.274G>A p.D92N Chr.11: 32097163-32097163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746580548 Chr.11: 32097098-32097098 on Build GRCh38
rs370966540 Chr.11: 32097119-32097119 on Build GRCh38
rs748722575 Chr.11: 32097138-32097138 on Build GRCh38
rs990834368 Chr.11: 32097026-32097026 on Build GRCh38
rs914738249 Chr.11: 32097065-32097065 on Build GRCh38
rs761887804 Chr.11: 32097158-32097158 on Build GRCh38
rs955001668 Chr.11: 32096987-32096987 on Build GRCh38
rs200171377 Chr.11: 32097141-32097141 on Build GRCh38
rs766033178 Chr.11: 32097171-32097171 on Build GRCh38
rs1031770497 Chr.11: 32096982-32096982 on Build GRCh38
rs974908849 Chr.11: 32097115-32097115 on Build GRCh38
rs374577571 Chr.11: 32097119-32097120 on Build GRCh38
rs202228198 Chr.11: 32097134-32097134 on Build GRCh38
rs963823978 Chr.11: 32097012-32097012 on Build GRCh38
rs144307253 Chr.11: 32097163-32097163 on Build GRCh38
rs758221377 Chr.11: 32097167-32097167 on Build GRCh38
rs201000684 Chr.11: 32097151-32097151 on Build GRCh38
rs778124672 Chr.11: 32097120-32097122 on Build GRCh38
rs748428306 Chr.11: 32097120-32097121 on Build GRCh38
rs910963399 Chr.11: 32097003-32097003 on Build GRCh38
rs184039815 Chr.11: 32097152-32097152 on Build GRCh38
rs986491128 Chr.11: 32097000-32097000 on Build GRCh38
rs556934664 Chr.11: 32097006-32097006 on Build GRCh38
rs756768072 Chr.11: 32096994-32096994 on Build GRCh38
rs564799768 Chr.11: 32097120-32097120 on Build GRCh38
rs199769797 Chr.11: 32097148-32097148 on Build GRCh38
rs77650900 Chr.11: 32097023-32097023 on Build GRCh38
rs943556806 Chr.11: 32097091-32097091 on Build GRCh38
rs373961205 Chr.11: 32097135-32097135 on Build GRCh38
rs771282884 Chr.11: 32097134-32097135 on Build GRCh38
rs754498680 Chr.11: 32097135-32097135 on Build GRCh38
rs925104131 Chr.11: 32097031-32097031 on Build GRCh38
rs138166885 Chr.11: 32097142-32097142 on Build GRCh38
rs191731908 Chr.11: 32096966-32096966 on Build GRCh38
rs149587906 Chr.11: 32097162-32097162 on Build GRCh38
rs767768797 Chr.11: 32097157-32097157 on Build GRCh38
rs780667169 Chr.11: 32097137-32097137 on Build GRCh38

View Full Product Details