Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 649586-649823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR90
Entrez Gene ID
197335
Gene Name
WD repeat domain 90
Gene Aliases
C16orf15, C16orf16, C16orf17, C16orf18, C16orf19
UniGene
Hs.511903
Gene Chromosome Location
Chr.16: 649363-667833 on Build GRCh38
Transcripts
NM_145294.4 XM_017023023.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6377261 c.16C>T p.Q6* Chr.16: 649768-649768 on Build GRCh38
4927808 c.70T>C p.S24P Chr.16: 649822-649822 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1050948324 Chr.16: 649604-649604 on Build GRCh38
rs753957135 Chr.16: 649802-649802 on Build GRCh38
rs1015661260 Chr.16: 649658-649658 on Build GRCh38
rs764019252 Chr.16: 649785-649785 on Build GRCh38
rs776217990 Chr.16: 649775-649775 on Build GRCh38
rs779864147 Chr.16: 649776-649776 on Build GRCh38
rs969784280 Chr.16: 649744-649744 on Build GRCh38
rs1004167265 Chr.16: 649645-649645 on Build GRCh38
rs1050241152 Chr.16: 649675-649675 on Build GRCh38
rs577818981 Chr.16: 649700-649700 on Build GRCh38
rs751393450 Chr.16: 649787-649787 on Build GRCh38
rs994459521 Chr.16: 649764-649764 on Build GRCh38
rs116801785 Chr.16: 649667-649667 on Build GRCh38
rs758900382 Chr.16: 649779-649779 on Build GRCh38
rs544940495 Chr.16: 649708-649708 on Build GRCh38
rs752528217 Chr.16: 649782-649782 on Build GRCh38
rs760361630 Chr.16: 649758-649758 on Build GRCh38
rs780082479 Chr.16: 649801-649801 on Build GRCh38
rs866493207 Chr.16: 649800-649800 on Build GRCh38
rs1038401195 Chr.16: 649711-649711 on Build GRCh38
rs537937084 Chr.16: 649636-649636 on Build GRCh38
rs773544773 Chr.16: 649751-649751 on Build GRCh38
rs770555703 Chr.16: 649762-649762 on Build GRCh38
rs762960909 Chr.16: 649784-649784 on Build GRCh38
rs765247122 Chr.16: 649780-649780 on Build GRCh38
rs911326434 Chr.16: 649698-649698 on Build GRCh38
rs772379995 Chr.16: 649740-649740 on Build GRCh38
rs981346147 Chr.16: 649746-649746 on Build GRCh38
rs767098252 Chr.16: 649631-649632 on Build GRCh38
rs915132850 Chr.16: 649617-649617 on Build GRCh38
rs1022635795 Chr.16: 649736-649736 on Build GRCh38
rs756464901 Chr.16: 649788-649788 on Build GRCh38
rs1035230667 Chr.16: 649747-649747 on Build GRCh38

View Full Product Details