Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54986280-54986531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM61
Entrez Gene ID
199964
Gene Name
transmembrane protein 61
Gene Aliases
-
UniGene
Hs.663950
Gene Chromosome Location
Chr.1: 54980614-54992293 on Build GRCh38
Transcripts
NM_182532.2 XM_005270586.4 XM_011540911.2 XM_011540912.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5352643 c.325T>C p.Y109H Chr.1: 54986406-54986406 on Build GRCh38
6879494 c.216C>T p.V72V Chr.1: 54986297-54986297 on Build GRCh38
2194271 c.234C>T p.G78G Chr.1: 54986315-54986315 on Build GRCh38
7183002 c.333C>T p.L111L Chr.1: 54986414-54986414 on Build GRCh38
910970 c.334A>C p.T112P Chr.1: 54986415-54986415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779644267 Chr.1: 54986423-54986423 on Build GRCh38
rs150272316 Chr.1: 54986372-54986372 on Build GRCh38
rs756070922 Chr.1: 54986492-54986492 on Build GRCh38
rs750277889 Chr.1: 54986403-54986403 on Build GRCh38
rs1055027557 Chr.1: 54986370-54986370 on Build GRCh38
rs200736187 Chr.1: 54986346-54986346 on Build GRCh38
rs144725989 Chr.1: 54986379-54986379 on Build GRCh38
rs769436044 Chr.1: 54986309-54986309 on Build GRCh38
rs761334255 Chr.1: 54986485-54986485 on Build GRCh38
rs779249058 Chr.1: 54986443-54986443 on Build GRCh38
rs1043361252 Chr.1: 54986342-54986342 on Build GRCh38
rs189457277 Chr.1: 54986507-54986507 on Build GRCh38
rs111620666 Chr.1: 54986471-54986471 on Build GRCh38
rs763774776 Chr.1: 54986478-54986478 on Build GRCh38
rs1003775180 Chr.1: 54986369-54986369 on Build GRCh38
rs766324136 Chr.1: 54986493-54986493 on Build GRCh38
rs369634808 Chr.1: 54986373-54986373 on Build GRCh38
rs750375288 Chr.1: 54986489-54986489 on Build GRCh38
rs373453802 Chr.1: 54986427-54986427 on Build GRCh38
rs745695557 Chr.1: 54986466-54986466 on Build GRCh38
rs199840145 Chr.1: 54986405-54986405 on Build GRCh38
rs56889206 Chr.1: 54986363-54986363 on Build GRCh38
rs756726906 Chr.1: 54986388-54986388 on Build GRCh38
rs967626772 Chr.1: 54986490-54986490 on Build GRCh38
rs777200745 Chr.1: 54986299-54986299 on Build GRCh38
rs749102837 Chr.1: 54986426-54986426 on Build GRCh38
rs371111022 Chr.1: 54986458-54986458 on Build GRCh38
rs184575542 Chr.1: 54986457-54986457 on Build GRCh38
rs761965433 Chr.1: 54986348-54986348 on Build GRCh38
rs745715296 Chr.1: 54986307-54986307 on Build GRCh38
rs550838536 Chr.1: 54986303-54986303 on Build GRCh38
rs200476053 Chr.1: 54986460-54986460 on Build GRCh38
rs267598658 Chr.1: 54986361-54986361 on Build GRCh38
rs755947686 Chr.1: 54986422-54986422 on Build GRCh38
rs372167123 Chr.1: 54986366-54986366 on Build GRCh38
rs773953280 Chr.1: 54986484-54986484 on Build GRCh38
rs138891775 Chr.1: 54986374-54986374 on Build GRCh38
rs199827287 Chr.1: 54986380-54986380 on Build GRCh38
rs142661965 Chr.1: 54986304-54986304 on Build GRCh38
rs748271782 Chr.1: 54986447-54986447 on Build GRCh38
rs779030670 Chr.1: 54986385-54986385 on Build GRCh38
rs952901083 Chr.1: 54986442-54986442 on Build GRCh38
rs780341647 Chr.1: 54986314-54986314 on Build GRCh38
rs367544152 Chr.1: 54986316-54986316 on Build GRCh38
rs767473897 Chr.1: 54986402-54986402 on Build GRCh38
rs199692872 Chr.1: 54986345-54986345 on Build GRCh38
rs201064478 Chr.1: 54986328-54986328 on Build GRCh38
rs766952665 Chr.1: 54986487-54986487 on Build GRCh38
rs113242933 Chr.1: 54986363-54986363 on Build GRCh38
rs542829571 Chr.1: 54986470-54986470 on Build GRCh38

View Full Product Details