Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 156693809-156694082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIPARP
Entrez Gene ID
25976
Gene Name
TCDD inducible poly(ADP-ribose) polymerase
Gene Aliases
ARTD14, PARP7, pART14
UniGene
Hs.744050
Gene Chromosome Location
Chr.3: 156674416-156706770 on Build GRCh38
Transcripts
NM_001184717.1 NM_001184718.1 NM_015508.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3427267 c.925G>T p.E309* Chr.3: 156694027-156694027 on Build GRCh38
1582566 c.964A>G p.T322A Chr.3: 156694066-156694066 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772729012 Chr.3: 156693972-156693972 on Build GRCh38
rs777458419 Chr.3: 156693999-156694002 on Build GRCh38
rs953292861 Chr.3: 156693996-156693996 on Build GRCh38
rs993054433 Chr.3: 156693879-156693879 on Build GRCh38
rs975753383 Chr.3: 156693993-156693993 on Build GRCh38
rs75767728 Chr.3: 156693920-156693920 on Build GRCh38
rs575941051 Chr.3: 156693861-156693861 on Build GRCh38
rs772228531 Chr.3: 156694009-156694009 on Build GRCh38
rs1006416380 Chr.3: 156693930-156693930 on Build GRCh38
rs1027349036 Chr.3: 156693846-156693846 on Build GRCh38
rs767912118 Chr.3: 156694002-156694002 on Build GRCh38
rs1024600299 Chr.3: 156693881-156693881 on Build GRCh38
rs7650182 Chr.3: 156693874-156693874 on Build GRCh38
rs746139475 Chr.3: 156694056-156694056 on Build GRCh38
rs765985945 Chr.3: 156693983-156693983 on Build GRCh38
rs193197 Chr.3: 156693943-156693943 on Build GRCh38
rs201812887 Chr.3: 156694015-156694015 on Build GRCh38
rs535410568 Chr.3: 156694008-156694009 on Build GRCh38
rs548113844 Chr.3: 156693944-156693944 on Build GRCh38
rs753095258 Chr.3: 156694010-156694010 on Build GRCh38
rs980294010 Chr.3: 156693888-156693888 on Build GRCh38
rs775746210 Chr.3: 156694018-156694018 on Build GRCh38
rs894740819 Chr.3: 156693891-156693891 on Build GRCh38
rs557662874 Chr.3: 156693859-156693859 on Build GRCh38
rs756568612 Chr.3: 156694013-156694013 on Build GRCh38
rs759878811 Chr.3: 156693997-156693997 on Build GRCh38
rs192753511 Chr.3: 156693862-156693862 on Build GRCh38
rs748090673 Chr.3: 156693994-156693994 on Build GRCh38
rs542589905 Chr.3: 156693837-156693837 on Build GRCh38
rs989309761 Chr.3: 156693945-156693945 on Build GRCh38
rs754480304 Chr.3: 156693993-156693994 on Build GRCh38
rs762379852 Chr.3: 156693982-156693982 on Build GRCh38
rs564445338 Chr.3: 156693873-156693873 on Build GRCh38
rs569582141 Chr.3: 156693964-156693964 on Build GRCh38
rs530642470 Chr.3: 156693970-156693970 on Build GRCh38
rs779184259 Chr.3: 156694023-156694023 on Build GRCh38
rs751109919 Chr.3: 156693990-156693991 on Build GRCh38
rs199900927 Chr.3: 156693994-156693994 on Build GRCh38
rs778160476 Chr.3: 156694014-156694014 on Build GRCh38
rs540275859 Chr.3: 156693882-156693882 on Build GRCh38
rs757348647 Chr.3: 156694020-156694020 on Build GRCh38
rs1013011743 Chr.3: 156693910-156693910 on Build GRCh38

View Full Product Details