Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 29528232-29528471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VSTM2B
Entrez Gene ID
342865
Gene Name
V-set and transmembrane domain containing 2B
Gene Aliases
-
UniGene
Hs.451618
Gene Chromosome Location
Chr.19: 29525431-29564555 on Build GRCh38
Transcripts
NM_001146339.1 XM_011526902.2 XM_011526903.2 XM_011526904.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7001634 c.297+2_297+3insT p.? Chr.19: 29528464-29528465 on Build GRCh38
6882428 c.297+1G>A p.? Chr.19: 29528463-29528463 on Build GRCh38
7332874 c.280G>T p.D94Y Chr.19: 29528445-29528445 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs538059362 Chr.19: 29528294-29528294 on Build GRCh38
rs980560634 Chr.19: 29528283-29528283 on Build GRCh38
rs992330528 Chr.19: 29528417-29528417 on Build GRCh38
rs931521188 Chr.19: 29528406-29528406 on Build GRCh38
rs577204897 Chr.19: 29528434-29528434 on Build GRCh38
rs913523993 Chr.19: 29528328-29528328 on Build GRCh38
rs368273940 Chr.19: 29528367-29528367 on Build GRCh38
rs945064416 Chr.19: 29528342-29528342 on Build GRCh38
rs1025077756 Chr.19: 29528280-29528280 on Build GRCh38
rs373947054 Chr.19: 29528405-29528405 on Build GRCh38
rs970506364 Chr.19: 29528281-29528281 on Build GRCh38
rs148923526 Chr.19: 29528442-29528442 on Build GRCh38
rs903838966 Chr.19: 29528373-29528373 on Build GRCh38
rs372018589 Chr.19: 29528397-29528397 on Build GRCh38
rs1048645149 Chr.19: 29528415-29528415 on Build GRCh38
rs1031086019 Chr.19: 29528396-29528396 on Build GRCh38
rs1004668365 Chr.19: 29528272-29528272 on Build GRCh38
rs781771898 Chr.19: 29528444-29528444 on Build GRCh38
rs1000981574 Chr.19: 29528389-29528389 on Build GRCh38
rs574792886 Chr.19: 29528366-29528366 on Build GRCh38
rs1027394144 Chr.19: 29528254-29528254 on Build GRCh38
rs1040843270 Chr.19: 29528255-29528255 on Build GRCh38
rs887086741 Chr.19: 29528423-29528423 on Build GRCh38
rs780369463 Chr.19: 29528361-29528361 on Build GRCh38
rs370561708 Chr.19: 29528426-29528426 on Build GRCh38
rs989445193 Chr.19: 29528290-29528290 on Build GRCh38
rs554857930 Chr.19: 29528341-29528341 on Build GRCh38
rs1025107093 Chr.19: 29528424-29528424 on Build GRCh38
rs919612975 Chr.19: 29528439-29528439 on Build GRCh38
rs987602148 Chr.19: 29528354-29528354 on Build GRCh38
rs1029429547 Chr.19: 29528363-29528363 on Build GRCh38
rs377593010 Chr.19: 29528284-29528284 on Build GRCh38

View Full Product Details