Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53237422-53237683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF677
Entrez Gene ID
342926
Gene Name
zinc finger protein 677
Gene Aliases
-
UniGene
Hs.20506
Gene Chromosome Location
Chr.19: 53235381-53254903 on Build GRCh38
Transcripts
NM_001317998.1 NM_182609.3 XM_005258862.4 XM_011526912.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5987466 c.1163C>G p.S388* Chr.19: 53237564-53237564 on Build GRCh38
6832330 c.1277C>A p.P426H Chr.19: 53237450-53237450 on Build GRCh38
6003881 c.1133A>G p.N378S Chr.19: 53237594-53237594 on Build GRCh38
6832331 c.1127A>C p.K376T Chr.19: 53237600-53237600 on Build GRCh38
6004320 c.1110T>A p.T370T Chr.19: 53237617-53237617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566157588 Chr.19: 53237547-53237548 on Build GRCh38
rs750611049 Chr.19: 53237549-53237549 on Build GRCh38
rs199860536 Chr.19: 53237484-53237484 on Build GRCh38
rs78900238 Chr.19: 53237471-53237471 on Build GRCh38
rs115065190 Chr.19: 53237528-53237528 on Build GRCh38
rs1001382439 Chr.19: 53237599-53237599 on Build GRCh38
rs756739916 Chr.19: 53237542-53237543 on Build GRCh38
rs748929742 Chr.19: 53237594-53237594 on Build GRCh38
rs750322607 Chr.19: 53237642-53237643 on Build GRCh38
rs769589020 Chr.19: 53237587-53237587 on Build GRCh38
rs369553139 Chr.19: 53237480-53237480 on Build GRCh38
rs781122431 Chr.19: 53237573-53237573 on Build GRCh38
rs773041798 Chr.19: 53237592-53237592 on Build GRCh38
rs761714870 Chr.19: 53237451-53237451 on Build GRCh38
rs867527371 Chr.19: 53237607-53237607 on Build GRCh38
rs777149195 Chr.19: 53237613-53237613 on Build GRCh38
rs762118209 Chr.19: 53237615-53237615 on Build GRCh38
rs770626831 Chr.19: 53237595-53237595 on Build GRCh38
rs371619640 Chr.19: 53237646-53237646 on Build GRCh38
rs769341998 Chr.19: 53237460-53237460 on Build GRCh38
rs759020869 Chr.19: 53237606-53237606 on Build GRCh38
rs757585609 Chr.19: 53237520-53237520 on Build GRCh38
rs949947772 Chr.19: 53237593-53237593 on Build GRCh38
rs780093697 Chr.19: 53237567-53237567 on Build GRCh38
rs925921133 Chr.19: 53237537-53237537 on Build GRCh38
rs867460164 Chr.19: 53237585-53237585 on Build GRCh38
rs144746199 Chr.19: 53237618-53237618 on Build GRCh38
rs750700545 Chr.19: 53237623-53237623 on Build GRCh38
rs147815623 Chr.19: 53237649-53237649 on Build GRCh38
rs936083763 Chr.19: 53237559-53237559 on Build GRCh38
rs751111637 Chr.19: 53237517-53237523 on Build GRCh38
rs763156280 Chr.19: 53237633-53237633 on Build GRCh38
rs781262411 Chr.19: 53237651-53237651 on Build GRCh38
rs142385940 Chr.19: 53237602-53237602 on Build GRCh38
rs201195757 Chr.19: 53237517-53237517 on Build GRCh38
rs201645076 Chr.19: 53237453-53237453 on Build GRCh38
rs754158475 Chr.19: 53237518-53237518 on Build GRCh38
rs752647244 Chr.19: 53237652-53237652 on Build GRCh38
rs769058077 Chr.19: 53237611-53237611 on Build GRCh38
rs866230433 Chr.19: 53237621-53237621 on Build GRCh38
rs147088386 Chr.19: 53237596-53237596 on Build GRCh38
rs772868349 Chr.19: 53237465-53237465 on Build GRCh38
rs8113436 Chr.19: 53237569-53237569 on Build GRCh38
rs565470305 Chr.19: 53237481-53237481 on Build GRCh38
rs747914929 Chr.19: 53237580-53237580 on Build GRCh38
rs747015213 Chr.19: 53237568-53237568 on Build GRCh38
rs1025434116 Chr.19: 53237650-53237650 on Build GRCh38
rs372722756 Chr.19: 53237506-53237506 on Build GRCh38
rs368860674 Chr.19: 53237571-53237571 on Build GRCh38
rs758520857 Chr.19: 53237562-53237562 on Build GRCh38
rs762580328 Chr.19: 53237477-53237477 on Build GRCh38

View Full Product Details