Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28673317-28673590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUPT6H
Entrez Gene ID
6830
Gene Name
SPT6 homolog, histone chaperone
Gene Aliases
SPT6, SPT6H, emb-5
UniGene
Hs.250429
Gene Chromosome Location
Chr.17: 28662183-28702684 on Build GRCh38
Transcripts
NM_001320755.1 NM_003170.4 XM_017024978.1 XM_017024979.1 XM_017024980.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
76636 c.52G>A p.D18N Chr.17: 28673453-28673453 on Build GRCh38
1381720 c.22G>A p.E8K Chr.17: 28673423-28673423 on Build GRCh38
417633 c.28G>A p.E10K Chr.17: 28673429-28673429 on Build GRCh38
4555832 c.66G>A p.V22V Chr.17: 28673467-28673467 on Build GRCh38
1193674 c.63G>T p.E21D Chr.17: 28673464-28673464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746505120 Chr.17: 28673475-28673475 on Build GRCh38
rs757743208 Chr.17: 28673396-28673396 on Build GRCh38
rs747778308 Chr.17: 28673464-28673464 on Build GRCh38
rs997599304 Chr.17: 28673426-28673426 on Build GRCh38
rs1011957525 Chr.17: 28673511-28673511 on Build GRCh38
rs370465811 Chr.17: 28673374-28673374 on Build GRCh38
rs781136616 Chr.17: 28673483-28673483 on Build GRCh38
rs964016462 Chr.17: 28673345-28673345 on Build GRCh38
rs769547362 Chr.17: 28673543-28673543 on Build GRCh38
rs759339518 Chr.17: 28673363-28673363 on Build GRCh38
rs756536549 Chr.17: 28673449-28673449 on Build GRCh38
rs9904043 Chr.17: 28673373-28673373 on Build GRCh38
rs985378271 Chr.17: 28673554-28673554 on Build GRCh38
rs764213903 Chr.17: 28673386-28673386 on Build GRCh38
rs767917683 Chr.17: 28673414-28673414 on Build GRCh38
rs375207346 Chr.17: 28673422-28673422 on Build GRCh38
rs758006296 Chr.17: 28673465-28673465 on Build GRCh38
rs896196015 Chr.17: 28673461-28673461 on Build GRCh38
rs751400299 Chr.17: 28673448-28673448 on Build GRCh38
rs887977770 Chr.17: 28673348-28673348 on Build GRCh38
rs369608990 Chr.17: 28673542-28673542 on Build GRCh38
rs897919382 Chr.17: 28673423-28673423 on Build GRCh38
rs770843147 Chr.17: 28673478-28673478 on Build GRCh38
rs953378527 Chr.17: 28673504-28673504 on Build GRCh38
rs191973369 Chr.17: 28673370-28673370 on Build GRCh38
rs780634959 Chr.17: 28673462-28673462 on Build GRCh38
rs777292621 Chr.17: 28673474-28673474 on Build GRCh38
rs776416712 Chr.17: 28673524-28673524 on Build GRCh38
rs775314476 Chr.17: 28673553-28673553 on Build GRCh38
rs1030771805 Chr.17: 28673477-28673477 on Build GRCh38
rs146417493 Chr.17: 28673355-28673355 on Build GRCh38
rs947626317 Chr.17: 28673360-28673360 on Build GRCh38
rs371900580 Chr.17: 28673394-28673394 on Build GRCh38
rs763113156 Chr.17: 28673560-28673560 on Build GRCh38
rs924822476 Chr.17: 28673383-28673383 on Build GRCh38
rs1056052725 Chr.17: 28673347-28673347 on Build GRCh38
rs1056222451 Chr.17: 28673389-28673389 on Build GRCh38

View Full Product Details