Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 10924419-10924692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XKR6
Entrez Gene ID
286046
Gene Name
XK related 6
Gene Aliases
C8orf21, C8orf5, C8orf7, XRG6
UniGene
Hs.657083
Gene Chromosome Location
Chr.8: 10896144-11205789 on Build GRCh38
Transcripts
NM_173683.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6823841 c.925A>G p.I309V Chr.8: 10924670-10924670 on Build GRCh38
3924465 c.960C>T p.P320P Chr.8: 10924635-10924635 on Build GRCh38
1270594 c.948C>T p.A316A Chr.8: 10924647-10924647 on Build GRCh38
118742 c.945C>T p.S315S Chr.8: 10924650-10924650 on Build GRCh38
3643856 c.939G>A p.K313K Chr.8: 10924656-10924656 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1052080545 Chr.8: 10924508-10924508 on Build GRCh38
rs190801580 Chr.8: 10924500-10924500 on Build GRCh38
rs982562203 Chr.8: 10924662-10924662 on Build GRCh38
rs772794829 Chr.8: 10924650-10924650 on Build GRCh38
rs546913542 Chr.8: 10924545-10924545 on Build GRCh38
rs752165884 Chr.8: 10924647-10924647 on Build GRCh38
rs899193886 Chr.8: 10924446-10924446 on Build GRCh38
rs996563164 Chr.8: 10924455-10924455 on Build GRCh38
rs1010617882 Chr.8: 10924528-10924528 on Build GRCh38
rs996205836 Chr.8: 10924563-10924563 on Build GRCh38
rs918594973 Chr.8: 10924479-10924479 on Build GRCh38
rs780144156 Chr.8: 10924624-10924624 on Build GRCh38
rs1029097803 Chr.8: 10924476-10924476 on Build GRCh38
rs753230172 Chr.8: 10924668-10924668 on Build GRCh38
rs529745391 Chr.8: 10924556-10924556 on Build GRCh38
rs571757055 Chr.8: 10924609-10924609 on Build GRCh38
rs371880369 Chr.8: 10924615-10924615 on Build GRCh38
rs1031092164 Chr.8: 10924485-10924485 on Build GRCh38
rs760183543 Chr.8: 10924652-10924652 on Build GRCh38
rs954930456 Chr.8: 10924478-10924478 on Build GRCh38
rs1022453677 Chr.8: 10924534-10924534 on Build GRCh38
rs762851086 Chr.8: 10924587-10924587 on Build GRCh38
rs761510924 Chr.8: 10924646-10924646 on Build GRCh38
rs7832003 Chr.8: 10924494-10924494 on Build GRCh38
rs202012600 Chr.8: 10924623-10924623 on Build GRCh38
rs970013938 Chr.8: 10924613-10924613 on Build GRCh38
rs763566122 Chr.8: 10924671-10924671 on Build GRCh38
rs758797685 Chr.8: 10924616-10924616 on Build GRCh38
rs530688645 Chr.8: 10924536-10924536 on Build GRCh38
rs956869294 Chr.8: 10924492-10924492 on Build GRCh38
rs560132179 Chr.8: 10924580-10924580 on Build GRCh38
rs369107658 Chr.8: 10924602-10924602 on Build GRCh38
rs768430213 Chr.8: 10924591-10924591 on Build GRCh38
rs376415583 Chr.8: 10924610-10924610 on Build GRCh38
rs368030191 Chr.8: 10924620-10924620 on Build GRCh38
rs150231499 Chr.8: 10924572-10924572 on Build GRCh38
rs371593422 Chr.8: 10924638-10924638 on Build GRCh38
rs561008584 Chr.8: 10924546-10924546 on Build GRCh38
rs969102993 Chr.8: 10924504-10924504 on Build GRCh38
rs765747323 Chr.8: 10924614-10924614 on Build GRCh38
rs552882213 Chr.8: 10924453-10924453 on Build GRCh38
rs778154948 Chr.8: 10924617-10924617 on Build GRCh38
rs1038313818 Chr.8: 10924557-10924557 on Build GRCh38
rs1031039807 Chr.8: 10924582-10924582 on Build GRCh38
rs745997950 Chr.8: 10924621-10924621 on Build GRCh38
rs899724681 Chr.8: 10924562-10924562 on Build GRCh38
rs567404688 Chr.8: 10924618-10924618 on Build GRCh38
rs980820168 Chr.8: 10924505-10924505 on Build GRCh38
rs749197966 Chr.8: 10924626-10924626 on Build GRCh38
rs572791567 Chr.8: 10924480-10924480 on Build GRCh38
rs1048959943 Chr.8: 10924482-10924482 on Build GRCh38
rs773907277 Chr.8: 10924601-10924601 on Build GRCh38
rs939048341 Chr.8: 10924517-10924517 on Build GRCh38
rs765839229 Chr.8: 10924659-10924659 on Build GRCh38
rs927747076 Chr.8: 10924511-10924511 on Build GRCh38
rs890544363 Chr.8: 10924573-10924573 on Build GRCh38
rs552004155 Chr.8: 10924604-10924604 on Build GRCh38

View Full Product Details