Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 43789627-43789874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRP1
Entrez Gene ID
8568
Gene Name
ribosomal RNA processing 1
Gene Aliases
D21S2056E, NNP-1, NOP52, RRP1A
UniGene
Hs.110757
Gene Chromosome Location
Chr.21: 43789537-43804102 on Build GRCh38
Transcripts
NM_003683.5 XM_017028485.1 XM_017028486.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6353656 c.49C>T p.L17L Chr.21: 43789678-43789678 on Build GRCh38
6398588 c.83C>T p.A28V Chr.21: 43789712-43789712 on Build GRCh38
4137268 c.2T>C p.M1T Chr.21: 43789631-43789631 on Build GRCh38
4650561 c.104A>G p.Y35C Chr.21: 43789733-43789733 on Build GRCh38
6559169 c.79C>T p.R27W Chr.21: 43789708-43789708 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1056404467 Chr.21: 43789770-43789770 on Build GRCh38
rs987108043 Chr.21: 43789817-43789817 on Build GRCh38
rs530432421 Chr.21: 43789694-43789694 on Build GRCh38
rs374138899 Chr.21: 43789849-43789849 on Build GRCh38
rs751501362 Chr.21: 43789744-43789744 on Build GRCh38
rs1044224753 Chr.21: 43789844-43789844 on Build GRCh38
rs1014136422 Chr.21: 43789703-43789703 on Build GRCh38
rs769307868 Chr.21: 43789686-43789686 on Build GRCh38
rs950463638 Chr.21: 43789786-43789786 on Build GRCh38
rs764821820 Chr.21: 43789802-43789802 on Build GRCh38
rs1024555666 Chr.21: 43789793-43789793 on Build GRCh38
rs759119620 Chr.21: 43789710-43789710 on Build GRCh38
rs912182823 Chr.21: 43789795-43789795 on Build GRCh38
rs879328209 Chr.21: 43789789-43789789 on Build GRCh38
rs756401494 Chr.21: 43789666-43789666 on Build GRCh38
rs757206842 Chr.21: 43789759-43789759 on Build GRCh38
rs983660138 Chr.21: 43789788-43789788 on Build GRCh38
rs373124510 Chr.21: 43789644-43789644 on Build GRCh38
rs746135596 Chr.21: 43789652-43789652 on Build GRCh38
rs923954289 Chr.21: 43789847-43789847 on Build GRCh38
rs78489660 Chr.21: 43789678-43789678 on Build GRCh38
rs767559581 Chr.21: 43789773-43789773 on Build GRCh38
rs966267369 Chr.21: 43789799-43789799 on Build GRCh38
rs774679026 Chr.21: 43789688-43789688 on Build GRCh38
rs963647895 Chr.21: 43789848-43789848 on Build GRCh38
rs547026072 Chr.21: 43789725-43789725 on Build GRCh38
rs533406291 Chr.21: 43789692-43789692 on Build GRCh38
rs762699166 Chr.21: 43789727-43789727 on Build GRCh38
rs969800443 Chr.21: 43789743-43789743 on Build GRCh38
rs770392911 Chr.21: 43789693-43789693 on Build GRCh38
rs199920102 Chr.21: 43789778-43789778 on Build GRCh38
rs548567025 Chr.21: 43789714-43789714 on Build GRCh38
rs946799862 Chr.21: 43789843-43789843 on Build GRCh38
rs866593435 Chr.21: 43789674-43789674 on Build GRCh38
rs749605541 Chr.21: 43789684-43789684 on Build GRCh38
rs973258588 Chr.21: 43789851-43789851 on Build GRCh38
rs763848237 Chr.21: 43789740-43789740 on Build GRCh38
rs778796403 Chr.21: 43789853-43789853 on Build GRCh38
rs2276245 Chr.21: 43789833-43789833 on Build GRCh38
rs1005178057 Chr.21: 43789816-43789816 on Build GRCh38

View Full Product Details