Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 104845776-104846049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC41A2
Entrez Gene ID
84102
Gene Name
solute carrier family 41 member 2
Gene Aliases
SLC41A1-L1
UniGene
Hs.577463
Gene Chromosome Location
Chr.12: 104802553-104958744 on Build GRCh38
Transcripts
NM_032148.3 XM_005269176.1 XM_005269178.1 XM_005269179.1 XM_006719630.2 XM_011538807.2 XM_011538808.1 XM_011538809.1 XM_011538810.2 XM_011538811.2 XM_011538813.2 XM_017020013.1 XM_017020014.1 XM_017020016.1 XM_017020017.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6782242 c.1319G>A p.S440N Chr.12: 104845911-104845911 on Build GRCh38
7093015 c.1268A>C p.N423T Chr.12: 104845962-104845962 on Build GRCh38
6782243 c.1070G>A p.S357N Chr.12: 104845911-104845911 on Build GRCh38
7093014 c.1019A>C p.N340T Chr.12: 104845962-104845962 on Build GRCh38
5701301 c.1285_1286insG p.A429fs*2 Chr.12: 104845944-104845945 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200609916 Chr.12: 104845977-104845977 on Build GRCh38
rs944960708 Chr.12: 104845896-104845896 on Build GRCh38
rs779698291 Chr.12: 104846018-104846018 on Build GRCh38
rs141246548 Chr.12: 104845942-104845942 on Build GRCh38
rs769665097 Chr.12: 104845897-104845897 on Build GRCh38
rs1038995571 Chr.12: 104845996-104845996 on Build GRCh38
rs746651487 Chr.12: 104845839-104845839 on Build GRCh38
rs760937335 Chr.12: 104845945-104845945 on Build GRCh38
rs554352457 Chr.12: 104845981-104845981 on Build GRCh38
rs912182403 Chr.12: 104845811-104845811 on Build GRCh38
rs776485499 Chr.12: 104845858-104845858 on Build GRCh38
rs772467806 Chr.12: 104846016-104846017 on Build GRCh38
rs78471645 Chr.12: 104846006-104846006 on Build GRCh38
rs373566107 Chr.12: 104845846-104845846 on Build GRCh38
rs372949695 Chr.12: 104845983-104845983 on Build GRCh38
rs985433643 Chr.12: 104845998-104845998 on Build GRCh38
rs759570351 Chr.12: 104845914-104845914 on Build GRCh38
rs764433019 Chr.12: 104845955-104845955 on Build GRCh38
rs748618080 Chr.12: 104845949-104845949 on Build GRCh38
rs376755765 Chr.12: 104845922-104845922 on Build GRCh38
rs761662638 Chr.12: 104845873-104845873 on Build GRCh38
rs779892475 Chr.12: 104845836-104845836 on Build GRCh38

View Full Product Details