Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 128795277-128795518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB7A
Entrez Gene ID
7879
Gene Name
RAB7A, member RAS oncogene family
Gene Aliases
PRO2706, RAB7
UniGene
Hs.684374
Gene Chromosome Location
Chr.3: 128726136-128814798 on Build GRCh38
Transcripts
NM_004637.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5708459 c.40C>G p.L14V Chr.3: 128795407-128795407 on Build GRCh38
3059611 c.46G>C p.D16H Chr.3: 128795413-128795413 on Build GRCh38
5604652 c.32T>C p.V11A Chr.3: 128795399-128795399 on Build GRCh38
6163271 c.27G>A p.L9L Chr.3: 128795394-128795394 on Build GRCh38
6841105 c.12_13insGAAGAAAGT p.R4_K5insEES Chr.3: 128795379-128795380 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1053225393 Chr.3: 128795373-128795373 on Build GRCh38
rs780732950 Chr.3: 128795401-128795401 on Build GRCh38
rs777086793 Chr.3: 128795362-128795362 on Build GRCh38
rs756651096 Chr.3: 128795394-128795394 on Build GRCh38
rs1000766383 Chr.3: 128795353-128795353 on Build GRCh38
rs752283088 Chr.3: 128795337-128795337 on Build GRCh38
rs932239784 Chr.3: 128795301-128795301 on Build GRCh38
rs771046614 Chr.3: 128795452-128795452 on Build GRCh38
rs372289310 Chr.3: 128795307-128795307 on Build GRCh38
rs748306081 Chr.3: 128795448-128795448 on Build GRCh38
rs570522911 Chr.3: 128795329-128795329 on Build GRCh38
rs115407768 Chr.3: 128795314-128795314 on Build GRCh38
rs774736035 Chr.3: 128795461-128795462 on Build GRCh38
rs150089960 Chr.3: 128795398-128795398 on Build GRCh38
rs772355381 Chr.3: 128795449-128795449 on Build GRCh38
rs766333135 Chr.3: 128795321-128795321 on Build GRCh38
rs760765904 Chr.3: 128795451-128795451 on Build GRCh38
rs377152050 Chr.3: 128795299-128795299 on Build GRCh38
rs147065732 Chr.3: 128795454-128795454 on Build GRCh38
rs759531766 Chr.3: 128795467-128795467 on Build GRCh38
rs764924961 Chr.3: 128795468-128795468 on Build GRCh38
rs760549551 Chr.3: 128795320-128795320 on Build GRCh38
rs149563047 Chr.3: 128795495-128795495 on Build GRCh38
rs182507583 Chr.3: 128795315-128795315 on Build GRCh38
rs187204546 Chr.3: 128795496-128795496 on Build GRCh38
rs140857794 Chr.3: 128795388-128795388 on Build GRCh38
rs753578967 Chr.3: 128795325-128795325 on Build GRCh38
rs752547155 Chr.3: 128795469-128795469 on Build GRCh38
rs749863950 Chr.3: 128795430-128795430 on Build GRCh38
rs768872098 Chr.3: 128795432-128795432 on Build GRCh38
rs779402683 Chr.3: 128795434-128795434 on Build GRCh38
rs368342555 Chr.3: 128795494-128795494 on Build GRCh38
rs758123417 Chr.3: 128795350-128795350 on Build GRCh38

View Full Product Details