Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68042928-68043183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCIRG1
Entrez Gene ID
10312
Gene Name
T-cell immune regulator 1, ATPase H+ transporting V0 subunit a3
Gene Aliases
ATP6N1C, ATP6V0A3, Atp6i, OC-116kDa, OC116, OPTB1, Stv1, TIRC7, Vph1, a3
UniGene
Hs.495985
Gene Chromosome Location
Chr.11: 68038995-68053839 on Build GRCh38
Transcripts
NM_006019.3 NM_006053.3 XM_005273709.3 XM_011544726.2 XM_017017089.1 XM_017017090.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6377206 c.454A>G p.T152A Chr.11: 68042982-68042982 on Build GRCh38
7247852 c.479G>A p.G160E Chr.11: 68043007-68043007 on Build GRCh38
6286934 c.418-9G>T p.? Chr.11: 68042937-68042937 on Build GRCh38
6384090 c.443C>T p.A148V Chr.11: 68042971-68042971 on Build GRCh38
5733799 c.482C>T p.P161L Chr.11: 68043010-68043010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573845313 Chr.11: 68043003-68043003 on Build GRCh38
rs925363548 Chr.11: 68042973-68042973 on Build GRCh38
rs199632510 Chr.11: 68043004-68043004 on Build GRCh38
rs749679691 Chr.11: 68043011-68043011 on Build GRCh38
rs780755928 Chr.11: 68042998-68042998 on Build GRCh38
rs774968760 Chr.11: 68043044-68043044 on Build GRCh38
rs371138836 Chr.11: 68042985-68042985 on Build GRCh38
rs754896714 Chr.11: 68042993-68042993 on Build GRCh38
rs34227834 Chr.11: 68043010-68043010 on Build GRCh38
rs1017915060 Chr.11: 68042958-68042958 on Build GRCh38
rs190476700 Chr.11: 68043093-68043093 on Build GRCh38
rs541001192 Chr.11: 68043141-68043141 on Build GRCh38
rs559475683 Chr.11: 68043165-68043165 on Build GRCh38
rs773690722 Chr.11: 68042959-68042959 on Build GRCh38
rs182676936 Chr.11: 68043115-68043115 on Build GRCh38
rs778942094 Chr.11: 68042998-68042998 on Build GRCh38
rs748413872 Chr.11: 68043082-68043082 on Build GRCh38
rs867889586 Chr.11: 68043148-68043148 on Build GRCh38
rs933494172 Chr.11: 68043068-68043068 on Build GRCh38
rs1006980323 Chr.11: 68042954-68042954 on Build GRCh38
rs575739241 Chr.11: 68043087-68043087 on Build GRCh38
rs761406373 Chr.11: 68042960-68042960 on Build GRCh38
rs888303395 Chr.11: 68043027-68043027 on Build GRCh38
rs986428885 Chr.11: 68043161-68043161 on Build GRCh38
rs750129963 Chr.11: 68042978-68042978 on Build GRCh38
rs753699657 Chr.11: 68042987-68042987 on Build GRCh38
rs898038011 Chr.11: 68043041-68043041 on Build GRCh38
rs772726088 Chr.11: 68043073-68043073 on Build GRCh38
rs1030559498 Chr.11: 68043147-68043147 on Build GRCh38
rs1001228717 Chr.11: 68043137-68043138 on Build GRCh38
rs746159308 Chr.11: 68043005-68043005 on Build GRCh38
rs761196309 Chr.11: 68043053-68043053 on Build GRCh38
rs892466007 Chr.11: 68042995-68042995 on Build GRCh38
rs186758849 Chr.11: 68043007-68043007 on Build GRCh38
rs766333949 Chr.11: 68042984-68042984 on Build GRCh38
rs979358405 Chr.11: 68042974-68042974 on Build GRCh38
rs767052351 Chr.11: 68042962-68042962 on Build GRCh38
rs965008893 Chr.11: 68043112-68043112 on Build GRCh38
rs751546005 Chr.11: 68043000-68043000 on Build GRCh38
rs536598236 Chr.11: 68043088-68043088 on Build GRCh38
rs949392573 Chr.11: 68043002-68043002 on Build GRCh38
rs370395754 Chr.11: 68043025-68043025 on Build GRCh38
rs771677132 Chr.11: 68043070-68043070 on Build GRCh38
rs1018385800 Chr.11: 68043113-68043113 on Build GRCh38
rs377425304 Chr.11: 68042955-68042955 on Build GRCh38
rs986723688 Chr.11: 68042964-68042964 on Build GRCh38
rs760540347 Chr.11: 68042983-68042983 on Build GRCh38

View Full Product Details