Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 117084456-117084711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAM1L1
Entrez Gene ID
133022
Gene Name
translocation associated membrane protein 1-like 1
Gene Aliases
-
UniGene
Hs.570737
Gene Chromosome Location
Chr.4: 117083554-117085580 on Build GRCh38
Transcripts
NM_152402.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1328720 c.905C>G p.S302W Chr.4: 117084489-117084489 on Build GRCh38
3365473 c.877G>T p.V293L Chr.4: 117084517-117084517 on Build GRCh38
6836394 c.730G>A p.E244K Chr.4: 117084664-117084664 on Build GRCh38
4584744 c.725G>A p.S242N Chr.4: 117084669-117084669 on Build GRCh38
4606638 c.730G>T p.E244* Chr.4: 117084664-117084664 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200676581 Chr.4: 117084671-117084671 on Build GRCh38
rs527665848 Chr.4: 117084568-117084568 on Build GRCh38
rs780613111 Chr.4: 117084679-117084679 on Build GRCh38
rs775345884 Chr.4: 117084518-117084518 on Build GRCh38
rs549346468 Chr.4: 117084575-117084575 on Build GRCh38
rs962128499 Chr.4: 117084673-117084673 on Build GRCh38
rs756351377 Chr.4: 117084548-117084548 on Build GRCh38
rs143062720 Chr.4: 117084536-117084536 on Build GRCh38
rs747412804 Chr.4: 117084682-117084682 on Build GRCh38
rs1015404475 Chr.4: 117084627-117084627 on Build GRCh38
rs982665500 Chr.4: 117084622-117084622 on Build GRCh38
rs142078992 Chr.4: 117084535-117084535 on Build GRCh38
rs760814608 Chr.4: 117084635-117084635 on Build GRCh38
rs772926593 Chr.4: 117084585-117084585 on Build GRCh38
rs374666796 Chr.4: 117084543-117084543 on Build GRCh38
rs769115461 Chr.4: 117084584-117084584 on Build GRCh38
rs754138688 Chr.4: 117084646-117084646 on Build GRCh38
rs757517960 Chr.4: 117084650-117084650 on Build GRCh38
rs562701564 Chr.4: 117084489-117084489 on Build GRCh38
rs368586988 Chr.4: 117084526-117084526 on Build GRCh38
rs778052836 Chr.4: 117084555-117084555 on Build GRCh38
rs868677235 Chr.4: 117084603-117084603 on Build GRCh38
rs1026899495 Chr.4: 117084564-117084564 on Build GRCh38
rs17852851 Chr.4: 117084662-117084662 on Build GRCh38
rs963691178 Chr.4: 117084521-117084521 on Build GRCh38
rs200297518 Chr.4: 117084544-117084544 on Build GRCh38
rs868126129 Chr.4: 117084596-117084599 on Build GRCh38
rs759517217 Chr.4: 117084593-117084593 on Build GRCh38
rs758892219 Chr.4: 117084676-117084676 on Build GRCh38
rs533288308 Chr.4: 117084524-117084524 on Build GRCh38
rs140073704 Chr.4: 117084590-117084590 on Build GRCh38
rs567559679 Chr.4: 117084638-117084638 on Build GRCh38
rs751520349 Chr.4: 117084528-117084528 on Build GRCh38
rs757713602 Chr.4: 117084561-117084561 on Build GRCh38
rs760513041 Chr.4: 117084557-117084561 on Build GRCh38
rs1006401887 Chr.4: 117084534-117084534 on Build GRCh38
rs764927567 Chr.4: 117084492-117084492 on Build GRCh38
rs767479036 Chr.4: 117084620-117084620 on Build GRCh38
rs749454354 Chr.4: 117084557-117084557 on Build GRCh38
rs747686922 Chr.4: 117084576-117084576 on Build GRCh38
rs752787981 Chr.4: 117084631-117084631 on Build GRCh38
rs768307094 Chr.4: 117084488-117084488 on Build GRCh38
rs779462778 Chr.4: 117084669-117084669 on Build GRCh38
rs762544664 Chr.4: 117084589-117084589 on Build GRCh38
rs147601937 Chr.4: 117084504-117084504 on Build GRCh38
rs151149444 Chr.4: 117084573-117084573 on Build GRCh38

View Full Product Details