Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 99613835-99614066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A28
Entrez Gene ID
81894
Gene Name
solute carrier family 25 member 28
Gene Aliases
MFRN2, MRS3/4, MRS4L, NPD016
UniGene
Hs.403790
Gene Chromosome Location
Chr.10: 99610518-99659329 on Build GRCh38
Transcripts
NM_031212.3 XM_005270208.1 XM_005270211.4 XM_006718005.3 XM_006718006.3 XM_011540239.2 XM_011540240.1 XM_011540244.2 XM_017016741.1 XM_017016742.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4429543 c.292-1G>T p.? Chr.10: 99613925-99613925 on Build GRCh38
6537102 c.355T>C p.W119R Chr.10: 99613861-99613861 on Build GRCh38
5369910 c.379C>T p.L127L Chr.10: 99613837-99613837 on Build GRCh38
4726838 c.371C>T p.T124M Chr.10: 99613845-99613845 on Build GRCh38
199799 c.335G>A p.R112H Chr.10: 99613881-99613881 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374194290 Chr.10: 99613948-99613948 on Build GRCh38
rs375360825 Chr.10: 99613881-99613881 on Build GRCh38
rs376744767 Chr.10: 99614023-99614024 on Build GRCh38
rs751800150 Chr.10: 99613907-99613907 on Build GRCh38
rs187023475 Chr.10: 99613973-99613973 on Build GRCh38
rs752839922 Chr.10: 99613882-99613882 on Build GRCh38
rs770467215 Chr.10: 99613947-99613947 on Build GRCh38
rs935305283 Chr.10: 99614002-99614002 on Build GRCh38
rs753846259 Chr.10: 99613874-99613874 on Build GRCh38
rs781499072 Chr.10: 99613920-99613920 on Build GRCh38
rs143249837 Chr.10: 99614008-99614008 on Build GRCh38
rs368020842 Chr.10: 99613883-99613883 on Build GRCh38
rs747721511 Chr.10: 99613963-99613963 on Build GRCh38
rs779559821 Chr.10: 99613979-99613979 on Build GRCh38
rs758605481 Chr.10: 99613887-99613887 on Build GRCh38
rs17494319 Chr.10: 99613972-99613972 on Build GRCh38
rs200683343 Chr.10: 99613888-99613888 on Build GRCh38
rs554700719 Chr.10: 99614006-99614006 on Build GRCh38
rs370300780 Chr.10: 99613865-99613865 on Build GRCh38
rs746352459 Chr.10: 99613932-99613932 on Build GRCh38
rs772873354 Chr.10: 99613861-99613861 on Build GRCh38

View Full Product Details