Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15848065-15848290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ37453
Entrez Gene ID
729614
Gene Name
uncharacterized LOC729614
Gene Aliases
-
UniGene
Hs.119298
Gene Chromosome Location
Chr.1: 15834215-15848147 on Build GRCh38
Transcripts
NR_024279.1 NM_015001.2

Gene Symbol
SPEN
Entrez Gene ID
23013
Gene Name
spen family transcriptional repressor
Gene Aliases
HIAA0929, MINT, RBM15C, SHARP
UniGene
Hs.744843
Gene Chromosome Location
Chr.1: 15847864-15940455 on Build GRCh38
Transcripts
NR_024279.1 NM_015001.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6959184 c.10G>A p.E4K Chr.1: 15848077-15848077 on Build GRCh38
6788658 c.53G>A p.R18Q Chr.1: 15848120-15848120 on Build GRCh38
6953253 c.55G>A p.E19K Chr.1: 15848122-15848122 on Build GRCh38
6915474 c.45G>T p.E15D Chr.1: 15848112-15848112 on Build GRCh38
4508881 c.78C>T p.F26F Chr.1: 15848145-15848145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750293261 Chr.1: 15848161-15848161 on Build GRCh38
rs776945585 Chr.1: 15848188-15848188 on Build GRCh38
rs755716858 Chr.1: 15848162-15848162 on Build GRCh38
rs566066982 Chr.1: 15848163-15848163 on Build GRCh38
rs552704761 Chr.1: 15848252-15848252 on Build GRCh38
rs148475723 Chr.1: 15848107-15848107 on Build GRCh38
rs754442199 Chr.1: 15848179-15848179 on Build GRCh38
rs757294150 Chr.1: 15848130-15848130 on Build GRCh38
rs751477102 Chr.1: 15848127-15848127 on Build GRCh38
rs775725887 Chr.1: 15848195-15848195 on Build GRCh38
rs747463297 Chr.1: 15848187-15848187 on Build GRCh38
rs199859041 Chr.1: 15848166-15848166 on Build GRCh38
rs868766295 Chr.1: 15848265-15848265 on Build GRCh38
rs202027212 Chr.1: 15848168-15848168 on Build GRCh38
rs551134634 Chr.1: 15848218-15848218 on Build GRCh38
rs778314062 Chr.1: 15848181-15848181 on Build GRCh38
rs537678178 Chr.1: 15848207-15848207 on Build GRCh38
rs1026772688 Chr.1: 15848217-15848217 on Build GRCh38
rs201094580 Chr.1: 15848192-15848192 on Build GRCh38
rs763092773 Chr.1: 15848196-15848196 on Build GRCh38
rs767372320 Chr.1: 15848160-15848160 on Build GRCh38
rs571106313 Chr.1: 15848220-15848220 on Build GRCh38
rs965200514 Chr.1: 15848255-15848255 on Build GRCh38
rs540116804 Chr.1: 15848244-15848244 on Build GRCh38
rs950728240 Chr.1: 15848244-15848246 on Build GRCh38

View Full Product Details