Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 177306365-177306638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MXD3
Entrez Gene ID
83463
Gene Name
MAX dimerization protein 3
Gene Aliases
BHLHC13, MAD3, MYX
UniGene
Hs.743332
Gene Chromosome Location
Chr.5: 177305500-177312291 on Build GRCh38
Transcripts
NM_001142935.1 NM_001271828.1 NM_013237.3

Gene Symbol
PRELID1
Entrez Gene ID
27166
Gene Name
PRELI domain containing 1
Gene Aliases
CGI-106, PRELI, PX19, SBBI12
UniGene
Hs.744904
Gene Chromosome Location
Chr.5: 177303762-177306959 on Build GRCh38
Transcripts
NM_001142935.1 NM_001271828.1 NM_013237.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
737588 c.594G>C p.E198D Chr.5: 177306504-177306504 on Build GRCh38
1066335 c.610G>A p.A204T Chr.5: 177306520-177306520 on Build GRCh38
3827769 c.514G>A p.E172K Chr.5: 177306424-177306424 on Build GRCh38
5762701 c.621G>A p.A207A Chr.5: 177306531-177306531 on Build GRCh38
5573165 c.512-7G>A p.? Chr.5: 177306415-177306415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11545838 Chr.5: 177306582-177306582 on Build GRCh38
rs199755338 Chr.5: 177306594-177306594 on Build GRCh38
rs780383596 Chr.5: 177306401-177306401 on Build GRCh38
rs775089583 Chr.5: 177306561-177306561 on Build GRCh38
rs765229378 Chr.5: 177306542-177306544 on Build GRCh38
rs760669214 Chr.5: 177306459-177306459 on Build GRCh38
rs764597274 Chr.5: 177306608-177306608 on Build GRCh38
rs374141318 Chr.5: 177306423-177306423 on Build GRCh38
rs891028469 Chr.5: 177306414-177306414 on Build GRCh38
rs9834 Chr.5: 177306615-177306615 on Build GRCh38
rs879053032 Chr.5: 177306427-177306427 on Build GRCh38
rs757559567 Chr.5: 177306539-177306539 on Build GRCh38
rs757111088 Chr.5: 177306571-177306571 on Build GRCh38
rs201616411 Chr.5: 177306531-177306531 on Build GRCh38
rs991651751 Chr.5: 177306560-177306560 on Build GRCh38
rs745664149 Chr.5: 177306614-177306614 on Build GRCh38
rs775331611 Chr.5: 177306436-177306436 on Build GRCh38
rs767581909 Chr.5: 177306597-177306597 on Build GRCh38
rs775781630 Chr.5: 177306471-177306471 on Build GRCh38
rs758980479 Chr.5: 177306424-177306424 on Build GRCh38
rs994470215 Chr.5: 177306478-177306478 on Build GRCh38
rs770853591 Chr.5: 177306485-177306485 on Build GRCh38
rs757628056 Chr.5: 177306617-177306617 on Build GRCh38
rs778485653 Chr.5: 177306544-177306544 on Build GRCh38
rs531296485 Chr.5: 177306410-177306410 on Build GRCh38
rs370214695 Chr.5: 177306415-177306415 on Build GRCh38
rs760242167 Chr.5: 177306390-177306391 on Build GRCh38
rs750724590 Chr.5: 177306396-177306396 on Build GRCh38
rs752842665 Chr.5: 177306542-177306547 on Build GRCh38
rs766257057 Chr.5: 177306394-177306395 on Build GRCh38
rs776656166 Chr.5: 177306567-177306567 on Build GRCh38
rs753521647 Chr.5: 177306415-177306416 on Build GRCh38
rs748681104 Chr.5: 177306411-177306411 on Build GRCh38
rs751795624 Chr.5: 177306501-177306501 on Build GRCh38
rs146349826 Chr.5: 177306486-177306486 on Build GRCh38
rs749656878 Chr.5: 177306536-177306536 on Build GRCh38
rs755083188 Chr.5: 177306506-177306506 on Build GRCh38
rs745528950 Chr.5: 177306550-177306550 on Build GRCh38
rs746478835 Chr.5: 177306562-177306562 on Build GRCh38
rs752801287 Chr.5: 177306598-177306598 on Build GRCh38
rs759779349 Chr.5: 177306585-177306585 on Build GRCh38
rs751599588 Chr.5: 177306404-177306404 on Build GRCh38
rs778084573 Chr.5: 177306417-177306417 on Build GRCh38
rs963873066 Chr.5: 177306555-177306555 on Build GRCh38
rs766528102 Chr.5: 177306492-177306492 on Build GRCh38
rs141608165 Chr.5: 177306563-177306563 on Build GRCh38
rs561502395 Chr.5: 177306421-177306421 on Build GRCh38
rs764074496 Chr.5: 177306462-177306462 on Build GRCh38
rs756632017 Chr.5: 177306527-177306527 on Build GRCh38
rs771597191 Chr.5: 177306435-177306435 on Build GRCh38
rs765079387 Chr.5: 177306580-177306580 on Build GRCh38
rs111655533 Chr.5: 177306519-177306519 on Build GRCh38
rs144966419 Chr.5: 177306522-177306522 on Build GRCh38
rs771533172 Chr.5: 177306554-177306554 on Build GRCh38
rs758716514 Chr.5: 177306400-177306400 on Build GRCh38
rs759554684 Chr.5: 177306533-177306535 on Build GRCh38
rs774474179 Chr.5: 177306583-177306583 on Build GRCh38

View Full Product Details