Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101039084-101039317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SFXN3
Entrez Gene ID
81855
Gene Name
sideroflexin 3
Gene Aliases
BA108L7.2, SFX3
UniGene
Hs.283844
Gene Chromosome Location
Chr.10: 101031229-101041243 on Build GRCh38
Transcripts
NM_030971.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6350769 c.864C>A p.A288A Chr.10: 101039205-101039205 on Build GRCh38
2058477 c.848delC p.L285fs*14 Chr.10: 101039189-101039189 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754053287 Chr.10: 101039172-101039172 on Build GRCh38
rs765632890 Chr.10: 101039166-101039166 on Build GRCh38
rs944240667 Chr.10: 101039260-101039260 on Build GRCh38
rs768361338 Chr.10: 101039243-101039243 on Build GRCh38
rs2277260 Chr.10: 101039205-101039205 on Build GRCh38
rs753501634 Chr.10: 101039247-101039247 on Build GRCh38
rs928610918 Chr.10: 101039151-101039151 on Build GRCh38
rs767522428 Chr.10: 101039129-101039129 on Build GRCh38
rs1033120934 Chr.10: 101039179-101039179 on Build GRCh38
rs917538442 Chr.10: 101039229-101039229 on Build GRCh38
rs769225089 Chr.10: 101039258-101039258 on Build GRCh38
rs372267494 Chr.10: 101039245-101039245 on Build GRCh38
rs772675968 Chr.10: 101039209-101039209 on Build GRCh38
rs778121482 Chr.10: 101039211-101039211 on Build GRCh38
rs762641333 Chr.10: 101039240-101039240 on Build GRCh38
rs750310791 Chr.10: 101039248-101039248 on Build GRCh38
rs760809183 Chr.10: 101039269-101039269 on Build GRCh38
rs774152297 Chr.10: 101039244-101039244 on Build GRCh38
rs887385050 Chr.10: 101039223-101039223 on Build GRCh38
rs543289710 Chr.10: 101039215-101039215 on Build GRCh38
rs769426357 Chr.10: 101039221-101039221 on Build GRCh38
rs557321224 Chr.10: 101039136-101039136 on Build GRCh38
rs991552697 Chr.10: 101039195-101039195 on Build GRCh38
rs775289139 Chr.10: 101039236-101039236 on Build GRCh38
rs756162029 Chr.10: 101039158-101039158 on Build GRCh38
rs780309391 Chr.10: 101039162-101039162 on Build GRCh38
rs958890827 Chr.10: 101039190-101039190 on Build GRCh38
rs376779961 Chr.10: 101039272-101039272 on Build GRCh38
rs545212828 Chr.10: 101039106-101039106 on Build GRCh38
rs145856296 Chr.10: 101039178-101039178 on Build GRCh38
rs779348187 Chr.10: 101039282-101039282 on Build GRCh38

View Full Product Details