Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99572609-99572870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF655
Entrez Gene ID
79027
Gene Name
zinc finger protein 655
Gene Aliases
VIK, VIK-1
UniGene
Hs.599798
Gene Chromosome Location
Chr.7: 99557726-99576453 on Build GRCh38
Transcripts
NM_001009960.1 NM_001083956.1 NM_001085368.1 NM_138494.2 XM_017012598.1 XM_017012599.1 XM_017012600.1 XM_017012601.1 XM_017012602.1 XM_017012603.1 XM_017012604.1 XM_017012605.1 XM_017012606.1 XM_017012607.1 XM_017012608.1 XM_017012609.1 XM_017012610.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
485808 c.555G>A p.Q185Q Chr.7: 99572663-99572663 on Build GRCh38
3367096 c.535A>T p.N179Y Chr.7: 99572643-99572643 on Build GRCh38
3663436 c.646A>G p.T216A Chr.7: 99572649-99572649 on Build GRCh38
4778512 c.556A>G p.S186G Chr.7: 99572664-99572664 on Build GRCh38
296886 c.513C>A p.A171A Chr.7: 99572621-99572621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374936662 Chr.7: 99572684-99572684 on Build GRCh38
rs774037605 Chr.7: 99572787-99572787 on Build GRCh38
rs1273679 Chr.7: 99572715-99572715 on Build GRCh38
rs748606200 Chr.7: 99572717-99572717 on Build GRCh38
rs752626820 Chr.7: 99572826-99572826 on Build GRCh38
rs752646892 Chr.7: 99572644-99572644 on Build GRCh38
rs774684468 Chr.7: 99572655-99572655 on Build GRCh38
rs747065673 Chr.7: 99572700-99572700 on Build GRCh38
rs767022292 Chr.7: 99572811-99572811 on Build GRCh38
rs1021431679 Chr.7: 99572841-99572841 on Build GRCh38
rs200453438 Chr.7: 99572672-99572672 on Build GRCh38
rs140597113 Chr.7: 99572800-99572800 on Build GRCh38
rs780463360 Chr.7: 99572688-99572688 on Build GRCh38
rs760557024 Chr.7: 99572832-99572832 on Build GRCh38
rs996451920 Chr.7: 99572677-99572677 on Build GRCh38
rs143155031 Chr.7: 99572836-99572836 on Build GRCh38
rs755874656 Chr.7: 99572645-99572645 on Build GRCh38
rs770535585 Chr.7: 99572783-99572783 on Build GRCh38
rs754859040 Chr.7: 99572651-99572652 on Build GRCh38
rs533962286 Chr.7: 99572632-99572632 on Build GRCh38
rs773470301 Chr.7: 99572740-99572740 on Build GRCh38
rs768917371 Chr.7: 99572711-99572711 on Build GRCh38
rs369268619 Chr.7: 99572737-99572737 on Build GRCh38
rs762622096 Chr.7: 99572759-99572759 on Build GRCh38
rs530882134 Chr.7: 99572784-99572784 on Build GRCh38

View Full Product Details