Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 116917866-116918093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFX6
Entrez Gene ID
222546
Gene Name
regulatory factor X6
Gene Aliases
MTCHRS, MTFS, RFXDC1, dJ955L16.1
UniGene
Hs.352276
Gene Chromosome Location
Chr.6: 116877213-116932163 on Build GRCh38
Transcripts
NM_173560.3 XM_011535589.1 XM_017010477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4185581 c.985G>A p.V329I Chr.6: 116918049-116918049 on Build GRCh38
6172053 c.997G>T p.A333S Chr.6: 116918061-116918061 on Build GRCh38
7251690 c.1022+5G>T p.? Chr.6: 116918091-116918091 on Build GRCh38
5413892 c.994C>T p.P332S Chr.6: 116918058-116918058 on Build GRCh38
4616921 c.1014G>T p.M338I Chr.6: 116918078-116918078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs957376159 Chr.6: 116918024-116918024 on Build GRCh38
rs760970395 Chr.6: 116918044-116918044 on Build GRCh38
rs6936629 Chr.6: 116917978-116917978 on Build GRCh38
rs369387201 Chr.6: 116918006-116918007 on Build GRCh38
rs763235902 Chr.6: 116917986-116917986 on Build GRCh38
rs1024924204 Chr.6: 116918029-116918029 on Build GRCh38
rs1014341297 Chr.6: 116917956-116917956 on Build GRCh38
rs903963108 Chr.6: 116917988-116917988 on Build GRCh38
rs753085200 Chr.6: 116918042-116918042 on Build GRCh38
rs894680341 Chr.6: 116917954-116917954 on Build GRCh38
rs1000971758 Chr.6: 116917992-116917992 on Build GRCh38
rs763668792 Chr.6: 116917953-116917953 on Build GRCh38
rs139343836 Chr.6: 116918049-116918049 on Build GRCh38
rs774468468 Chr.6: 116918000-116918000 on Build GRCh38
rs1054799884 Chr.6: 116917936-116917936 on Build GRCh38
rs367721560 Chr.6: 116918010-116918010 on Build GRCh38
rs902088579 Chr.6: 116917961-116917961 on Build GRCh38
rs1007690384 Chr.6: 116918026-116918026 on Build GRCh38
rs13213246 Chr.6: 116917923-116917923 on Build GRCh38
rs200963523 Chr.6: 116918001-116918002 on Build GRCh38
rs201669550 Chr.6: 116918007-116918007 on Build GRCh38
rs371078050 Chr.6: 116917987-116917987 on Build GRCh38
rs549113998 Chr.6: 116917925-116917925 on Build GRCh38

View Full Product Details