Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149309937-149310206 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFAP1L1
Entrez Gene ID
134265
Gene Name
actin filament associated protein 1 like 1
Gene Aliases
-
UniGene
Hs.483793
Gene Chromosome Location
Chr.5: 149271838-149343637 on Build GRCh38
Transcripts
NM_001146337.2 NM_001323062.1 NM_001323063.1 NM_152406.3 XM_011537558.1 XM_017009036.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6448435 c.796G>A p.D266N Chr.5: 149310004-149310004 on Build GRCh38
6535775 c.828C>G p.D276E Chr.5: 149310036-149310036 on Build GRCh38
6414762 c.862C>T p.Q288* Chr.5: 149310070-149310070 on Build GRCh38
1064140 c.854G>A p.R285H Chr.5: 149310062-149310062 on Build GRCh38
6595747 c.832C>T p.R278W Chr.5: 149310040-149310040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890715438 Chr.5: 149310117-149310117 on Build GRCh38
rs772429268 Chr.5: 149310028-149310028 on Build GRCh38
rs142531986 Chr.5: 149309978-149309978 on Build GRCh38
rs779849365 Chr.5: 149310173-149310173 on Build GRCh38
rs765391733 Chr.5: 149310047-149310047 on Build GRCh38
rs764018151 Chr.5: 149310069-149310069 on Build GRCh38
rs373991708 Chr.5: 149309977-149309977 on Build GRCh38
rs773435877 Chr.5: 149310051-149310051 on Build GRCh38
rs751317092 Chr.5: 149309994-149309994 on Build GRCh38
rs375382983 Chr.5: 149310154-149310154 on Build GRCh38
rs149361551 Chr.5: 149310075-149310075 on Build GRCh38
rs368271604 Chr.5: 149310040-149310040 on Build GRCh38
rs749881630 Chr.5: 149310161-149310161 on Build GRCh38
rs375742704 Chr.5: 149310062-149310062 on Build GRCh38
rs202169673 Chr.5: 149310027-149310027 on Build GRCh38
rs536630154 Chr.5: 149310113-149310113 on Build GRCh38
rs775889847 Chr.5: 149310030-149310030 on Build GRCh38
rs1020424330 Chr.5: 149310148-149310148 on Build GRCh38
rs767379415 Chr.5: 149310008-149310008 on Build GRCh38
rs993294784 Chr.5: 149309989-149309989 on Build GRCh38
rs760672570 Chr.5: 149310126-149310126 on Build GRCh38
rs144677131 Chr.5: 149310099-149310099 on Build GRCh38
rs139614291 Chr.5: 149310054-149310054 on Build GRCh38
rs771042750 Chr.5: 149310130-149310130 on Build GRCh38
rs753355489 Chr.5: 149310070-149310070 on Build GRCh38
rs767387289 Chr.5: 149310055-149310055 on Build GRCh38
rs534438478 Chr.5: 149310123-149310123 on Build GRCh38
rs192371998 Chr.5: 149310023-149310023 on Build GRCh38
rs555105102 Chr.5: 149310120-149310120 on Build GRCh38
rs756695294 Chr.5: 149310073-149310073 on Build GRCh38
rs762174067 Chr.5: 149309979-149309979 on Build GRCh38
rs747556491 Chr.5: 149310039-149310039 on Build GRCh38
rs567317757 Chr.5: 149310121-149310121 on Build GRCh38
rs766331502 Chr.5: 149310176-149310177 on Build GRCh38
rs1022489791 Chr.5: 149310183-149310183 on Build GRCh38
rs371084172 Chr.5: 149310061-149310061 on Build GRCh38
rs1000826402 Chr.5: 149310162-149310162 on Build GRCh38
rs765542480 Chr.5: 149309988-149309988 on Build GRCh38
rs34618588 Chr.5: 149310172-149310172 on Build GRCh38
rs61730364 Chr.5: 149310126-149310126 on Build GRCh38
rs757954923 Chr.5: 149310104-149310104 on Build GRCh38
rs761263807 Chr.5: 149310144-149310144 on Build GRCh38
rs777317173 Chr.5: 149310017-149310017 on Build GRCh38
rs748787542 Chr.5: 149310021-149310021 on Build GRCh38
rs751977804 Chr.5: 149310177-149310177 on Build GRCh38
rs148497345 Chr.5: 149310110-149310110 on Build GRCh38
rs755345391 Chr.5: 149310181-149310181 on Build GRCh38
rs964320149 Chr.5: 149310157-149310157 on Build GRCh38
rs776907700 Chr.5: 149310041-149310041 on Build GRCh38
rs35622916 Chr.5: 149310076-149310076 on Build GRCh38
rs760857095 Chr.5: 149309967-149309967 on Build GRCh38
rs760391981 Chr.5: 149310136-149310136 on Build GRCh38
rs146039286 Chr.5: 149310025-149310025 on Build GRCh38
rs764036594 Chr.5: 149310139-149310139 on Build GRCh38
rs752641464 Chr.5: 149310013-149310013 on Build GRCh38
rs781772197 Chr.5: 149310185-149310185 on Build GRCh38
rs199506796 Chr.5: 149309973-149309973 on Build GRCh38
rs763282756 Chr.5: 149310129-149310129 on Build GRCh38
rs754745832 Chr.5: 149310001-149310001 on Build GRCh38
rs761979787 Chr.5: 149310044-149310044 on Build GRCh38
rs778448869 Chr.5: 149310082-149310082 on Build GRCh38
rs753729323 Chr.5: 149310140-149310140 on Build GRCh38
rs372100607 Chr.5: 149310131-149310131 on Build GRCh38

View Full Product Details