Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74459546-74459819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INO80B-WBP1
Entrez Gene ID
100532735
Gene Name
INO80B-WBP1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.410786
Gene Chromosome Location
Chr.2: 74455023-74460891 on Build GRCh38
Transcripts
NR_037849.1 NM_012477.3

Gene Symbol
WBP1
Entrez Gene ID
23559
Gene Name
WW domain binding protein 1
Gene Aliases
WBP-1
UniGene
Hs.516114
Gene Chromosome Location
Chr.2: 74458400-74460891 on Build GRCh38
Transcripts
NR_037849.1 NM_012477.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3583363 c.171G>A p.W57* Chr.2: 74459744-74459744 on Build GRCh38
5484565 c.133G>A p.G45R Chr.2: 74459706-74459706 on Build GRCh38
4740770 c.166C>T p.L56F Chr.2: 74459739-74459739 on Build GRCh38
7397567 c.119G>A p.S40N Chr.2: 74459692-74459692 on Build GRCh38
6415118 c.142G>A p.G48S Chr.2: 74459715-74459715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751264132 Chr.2: 74459758-74459758 on Build GRCh38
rs773463304 Chr.2: 74459716-74459716 on Build GRCh38
rs138531286 Chr.2: 74459650-74459650 on Build GRCh38
rs369119714 Chr.2: 74459609-74459610 on Build GRCh38
rs779801549 Chr.2: 74459724-74459725 on Build GRCh38
rs144737803 Chr.2: 74459705-74459705 on Build GRCh38
rs199876747 Chr.2: 74459746-74459746 on Build GRCh38
rs201694119 Chr.2: 74459749-74459749 on Build GRCh38
rs986970087 Chr.2: 74459612-74459612 on Build GRCh38
rs765325474 Chr.2: 74459729-74459729 on Build GRCh38
rs199793567 Chr.2: 74459725-74459725 on Build GRCh38
rs749368738 Chr.2: 74459610-74459610 on Build GRCh38
rs771143913 Chr.2: 74459792-74459792 on Build GRCh38
rs932229455 Chr.2: 74459652-74459652 on Build GRCh38
rs748428409 Chr.2: 74459776-74459776 on Build GRCh38
rs371298640 Chr.2: 74459774-74459774 on Build GRCh38
rs749832174 Chr.2: 74459771-74459771 on Build GRCh38
rs772487221 Chr.2: 74459778-74459778 on Build GRCh38
rs922167829 Chr.2: 74459646-74459646 on Build GRCh38
rs755632193 Chr.2: 74459766-74459766 on Build GRCh38
rs774646070 Chr.2: 74459600-74459600 on Build GRCh38
rs779472222 Chr.2: 74459768-74459768 on Build GRCh38
rs760924629 Chr.2: 74459786-74459786 on Build GRCh38
rs779380136 Chr.2: 74459775-74459775 on Build GRCh38
rs966267090 Chr.2: 74459601-74459601 on Build GRCh38
rs772398178 Chr.2: 74459708-74459708 on Build GRCh38
rs767663157 Chr.2: 74459611-74459611 on Build GRCh38
rs771102692 Chr.2: 74459692-74459693 on Build GRCh38
rs775961992 Chr.2: 74459690-74459690 on Build GRCh38
rs764279579 Chr.2: 74459593-74459593 on Build GRCh38
rs1006534376 Chr.2: 74459738-74459738 on Build GRCh38
rs544990744 Chr.2: 74459667-74459667 on Build GRCh38
rs752609424 Chr.2: 74459639-74459639 on Build GRCh38
rs773557702 Chr.2: 74459783-74459783 on Build GRCh38
rs758201113 Chr.2: 74459649-74459649 on Build GRCh38
rs147307399 Chr.2: 74459699-74459699 on Build GRCh38
rs1049415878 Chr.2: 74459663-74459663 on Build GRCh38
rs1016202560 Chr.2: 74459751-74459751 on Build GRCh38
rs749991794 Chr.2: 74459762-74459762 on Build GRCh38
rs760836503 Chr.2: 74459720-74459720 on Build GRCh38
rs756985690 Chr.2: 74459760-74459760 on Build GRCh38
rs762971913 Chr.2: 74459794-74459794 on Build GRCh38
rs755012241 Chr.2: 74459631-74459631 on Build GRCh38
rs763981864 Chr.2: 74459757-74459757 on Build GRCh38
rs909585806 Chr.2: 74459681-74459681 on Build GRCh38
rs374169904 Chr.2: 74459678-74459678 on Build GRCh38
rs745361247 Chr.2: 74459679-74459679 on Build GRCh38
rs770326978 Chr.2: 74459688-74459688 on Build GRCh38
rs201891117 Chr.2: 74459756-74459756 on Build GRCh38
rs1023415075 Chr.2: 74459726-74459728 on Build GRCh38
rs148596865 Chr.2: 74459692-74459692 on Build GRCh38
rs780793659 Chr.2: 74459761-74459761 on Build GRCh38
rs191675891 Chr.2: 74459638-74459638 on Build GRCh38
rs769068059 Chr.2: 74459693-74459693 on Build GRCh38
rs201613495 Chr.2: 74459621-74459621 on Build GRCh38

View Full Product Details