Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 179121628-179121853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS2
Entrez Gene ID
9509
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 2
Gene Aliases
ADAM-TS2, ADAMTS-2, ADAMTS-3, NPI, PC I-NP, PCI-NP, PCINP, PCPNI, PNPI
UniGene
Hs.23871
Gene Chromosome Location
Chr.5: 179110851-179345430 on Build GRCh38
Transcripts
NM_014244.4 XM_017010063.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6594137 c.3120C>T p.Y1040Y Chr.5: 179121719-179121719 on Build GRCh38
6576653 c.3178+6C>T p.? Chr.5: 179121655-179121655 on Build GRCh38
333457 c.3121G>A p.V1041I Chr.5: 179121718-179121718 on Build GRCh38
6594142 c.3139C>T p.R1047C Chr.5: 179121700-179121700 on Build GRCh38
1436472 c.3173C>T p.S1058L Chr.5: 179121666-179121666 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888656746 Chr.5: 179121833-179121833 on Build GRCh38
rs756484519 Chr.5: 179121799-179121799 on Build GRCh38
rs781475895 Chr.5: 179121703-179121703 on Build GRCh38
rs1033066690 Chr.5: 179121832-179121832 on Build GRCh38
rs775155341 Chr.5: 179121669-179121669 on Build GRCh38
rs1028862410 Chr.5: 179121735-179121735 on Build GRCh38
rs948167718 Chr.5: 179121657-179121657 on Build GRCh38
rs535196543 Chr.5: 179121684-179121684 on Build GRCh38
rs763966576 Chr.5: 179121683-179121683 on Build GRCh38
rs750141517 Chr.5: 179121696-179121696 on Build GRCh38
rs779277426 Chr.5: 179121732-179121732 on Build GRCh38
rs201390756 Chr.5: 179121685-179121685 on Build GRCh38
rs888936111 Chr.5: 179121744-179121744 on Build GRCh38
rs913554063 Chr.5: 179121774-179121774 on Build GRCh38
rs977115988 Chr.5: 179121831-179121831 on Build GRCh38
rs775312777 Chr.5: 179121748-179121748 on Build GRCh38
rs957771345 Chr.5: 179121742-179121742 on Build GRCh38
rs879240385 Chr.5: 179121824-179121824 on Build GRCh38
rs753074786 Chr.5: 179121790-179121790 on Build GRCh38
rs778184902 Chr.5: 179121718-179121718 on Build GRCh38
rs760609413 Chr.5: 179121676-179121676 on Build GRCh38
rs774767519 Chr.5: 179121654-179121654 on Build GRCh38
rs765979484 Chr.5: 179121783-179121783 on Build GRCh38
rs374907597 Chr.5: 179121689-179121689 on Build GRCh38
rs922829240 Chr.5: 179121822-179121822 on Build GRCh38
rs967496242 Chr.5: 179121804-179121804 on Build GRCh38
rs1036733329 Chr.5: 179121695-179121695 on Build GRCh38
rs764660233 Chr.5: 179121771-179121771 on Build GRCh38
rs749365337 Chr.5: 179121719-179121719 on Build GRCh38
rs753027177 Chr.5: 179121704-179121704 on Build GRCh38
rs767490706 Chr.5: 179121666-179121666 on Build GRCh38
rs762546707 Chr.5: 179121782-179121782 on Build GRCh38
rs976464628 Chr.5: 179121812-179121813 on Build GRCh38
rs776485241 Chr.5: 179121759-179121759 on Build GRCh38
rs760424301 Chr.5: 179121757-179121757 on Build GRCh38
rs759395261 Chr.5: 179121656-179121656 on Build GRCh38
rs756604769 Chr.5: 179121709-179121709 on Build GRCh38
rs761307854 Chr.5: 179121769-179121769 on Build GRCh38
rs772438979 Chr.5: 179121745-179121745 on Build GRCh38
rs746024135 Chr.5: 179121741-179121741 on Build GRCh38
rs758048360 Chr.5: 179121700-179121700 on Build GRCh38
rs554723419 Chr.5: 179121775-179121775 on Build GRCh38
rs771197106 Chr.5: 179121723-179121723 on Build GRCh38
rs368918205 Chr.5: 179121797-179121797 on Build GRCh38
rs573137553 Chr.5: 179121823-179121823 on Build GRCh38

View Full Product Details