Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58357380-58357653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF43
Entrez Gene ID
54894
Gene Name
ring finger protein 43
Gene Aliases
RNF124, URCC
UniGene
Hs.584916
Gene Chromosome Location
Chr.17: 58352500-58418895 on Build GRCh38
Transcripts
NM_001305544.1 NM_001305545.1 NM_017763.5 XM_011524955.2 XM_011524956.2 XM_017024800.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4392568 c.2260G>C p.G754R Chr.17: 58357516-58357516 on Build GRCh38
1287672 c.2177G>T p.W726L Chr.17: 58357599-58357599 on Build GRCh38
5909622 c.2215C>T p.P739S Chr.17: 58357561-58357561 on Build GRCh38
1640760 c.2280G>A p.P760P Chr.17: 58357496-58357496 on Build GRCh38
6767979 c.2181G>A p.L727L Chr.17: 58357595-58357595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs969759215 Chr.17: 58357619-58357619 on Build GRCh38
rs772096609 Chr.17: 58357581-58357581 on Build GRCh38
rs770712571 Chr.17: 58357550-58357550 on Build GRCh38
rs759690943 Chr.17: 58357558-58357558 on Build GRCh38
rs775335997 Chr.17: 58357582-58357582 on Build GRCh38
rs750460220 Chr.17: 58357632-58357632 on Build GRCh38
rs773941703 Chr.17: 58357552-58357552 on Build GRCh38
rs375779794 Chr.17: 58357431-58357431 on Build GRCh38
rs765464591 Chr.17: 58357629-58357629 on Build GRCh38
rs564215300 Chr.17: 58357490-58357490 on Build GRCh38
rs780567055 Chr.17: 58357519-58357519 on Build GRCh38
rs891451977 Chr.17: 58357553-58357553 on Build GRCh38
rs777471441 Chr.17: 58357528-58357528 on Build GRCh38
rs1039051017 Chr.17: 58357547-58357547 on Build GRCh38
rs776489595 Chr.17: 58357628-58357628 on Build GRCh38
rs745395155 Chr.17: 58357576-58357576 on Build GRCh38
rs769450192 Chr.17: 58357471-58357471 on Build GRCh38
rs759480914 Chr.17: 58357492-58357492 on Build GRCh38
rs755350796 Chr.17: 58357523-58357523 on Build GRCh38
rs772783321 Chr.17: 58357479-58357479 on Build GRCh38
rs764354670 Chr.17: 58357503-58357503 on Build GRCh38
rs760426158 Chr.17: 58357497-58357497 on Build GRCh38
rs758906251 Chr.17: 58357426-58357426 on Build GRCh38
rs773107390 Chr.17: 58357592-58357592 on Build GRCh38
rs747371137 Chr.17: 58357522-58357522 on Build GRCh38
rs61746279 Chr.17: 58357496-58357496 on Build GRCh38
rs758850811 Chr.17: 58357517-58357517 on Build GRCh38
rs931342515 Chr.17: 58357473-58357473 on Build GRCh38
rs973765221 Chr.17: 58357451-58357451 on Build GRCh38
rs1047877953 Chr.17: 58357505-58357505 on Build GRCh38
rs115553539 Chr.17: 58357515-58357515 on Build GRCh38
rs766430982 Chr.17: 58357443-58357443 on Build GRCh38
rs754064531 Chr.17: 58357508-58357508 on Build GRCh38
rs199912588 Chr.17: 58357612-58357612 on Build GRCh38
rs748813654 Chr.17: 58357482-58357482 on Build GRCh38
rs748964171 Chr.17: 58357541-58357541 on Build GRCh38
rs767576953 Chr.17: 58357493-58357493 on Build GRCh38
rs757324156 Chr.17: 58357509-58357509 on Build GRCh38
rs973433777 Chr.17: 58357445-58357445 on Build GRCh38
rs765229920 Chr.17: 58357514-58357514 on Build GRCh38
rs143655541 Chr.17: 58357534-58357534 on Build GRCh38
rs760504747 Chr.17: 58357620-58357620 on Build GRCh38
rs370703233 Chr.17: 58357542-58357542 on Build GRCh38
rs780335907 Chr.17: 58357427-58357427 on Build GRCh38
rs768401917 Chr.17: 58357627-58357627 on Build GRCh38
rs746441025 Chr.17: 58357423-58357423 on Build GRCh38

View Full Product Details