Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96253833-96254070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM127
Entrez Gene ID
55654
Gene Name
transmembrane protein 127
Gene Aliases
-
UniGene
Hs.164303
Gene Chromosome Location
Chr.2: 96248514-96266014 on Build GRCh38
Transcripts
NM_001193304.2 NM_017849.3 XM_017004450.1 XM_017004451.1 XM_017004452.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4922910 c.459C>T p.L153L Chr.2: 96254066-96254066 on Build GRCh38
4935520 c.460A>G p.I154V Chr.2: 96254065-96254065 on Build GRCh38
1023855 c.523G>A p.V175I Chr.2: 96254002-96254002 on Build GRCh38
1023856 c.482_483insGCA p.Q160_H161insQ Chr.2: 96254042-96254043 on Build GRCh38
6978701 c.502C>T p.Q168* Chr.2: 96254023-96254023 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753594901 Chr.2: 96254005-96254005 on Build GRCh38
rs772481029 Chr.2: 96253859-96253859 on Build GRCh38
rs774413835 Chr.2: 96253971-96253971 on Build GRCh38
rs200327514 Chr.2: 96253953-96253953 on Build GRCh38
rs367951215 Chr.2: 96254003-96254003 on Build GRCh38
rs767190490 Chr.2: 96253915-96253915 on Build GRCh38
rs140860906 Chr.2: 96253936-96253936 on Build GRCh38
rs775177724 Chr.2: 96253891-96253891 on Build GRCh38
rs368792184 Chr.2: 96253892-96253892 on Build GRCh38
rs149034651 Chr.2: 96254002-96254002 on Build GRCh38
rs373781978 Chr.2: 96253957-96253957 on Build GRCh38
rs764012422 Chr.2: 96253969-96253969 on Build GRCh38
rs536032990 Chr.2: 96254039-96254039 on Build GRCh38
rs759801591 Chr.2: 96253874-96253874 on Build GRCh38
rs762657413 Chr.2: 96253963-96253963 on Build GRCh38
rs146965678 Chr.2: 96253960-96253960 on Build GRCh38
rs752030320 Chr.2: 96254031-96254031 on Build GRCh38
rs147816248 Chr.2: 96253952-96253952 on Build GRCh38
rs121908831 Chr.2: 96253884-96253885 on Build GRCh38
rs778839133 Chr.2: 96254043-96254043 on Build GRCh38
rs747098700 Chr.2: 96253861-96253861 on Build GRCh38
rs779910556 Chr.2: 96253858-96253858 on Build GRCh38
rs144326855 Chr.2: 96253863-96253863 on Build GRCh38
rs372120755 Chr.2: 96254029-96254029 on Build GRCh38
rs750072653 Chr.2: 96253916-96253916 on Build GRCh38
rs776732082 Chr.2: 96253958-96253958 on Build GRCh38
rs767100341 Chr.2: 96253987-96253987 on Build GRCh38
rs757792507 Chr.2: 96254036-96254036 on Build GRCh38
rs765050645 Chr.2: 96253884-96253884 on Build GRCh38
rs745947594 Chr.2: 96254040-96254040 on Build GRCh38
rs550833832 Chr.2: 96253991-96253991 on Build GRCh38
rs751044006 Chr.2: 96253905-96253905 on Build GRCh38
rs377760956 Chr.2: 96253862-96253862 on Build GRCh38
rs755757863 Chr.2: 96253919-96253919 on Build GRCh38
rs1033423510 Chr.2: 96254004-96254004 on Build GRCh38
rs758676810 Chr.2: 96254006-96254006 on Build GRCh38
rs758874572 Chr.2: 96253940-96253940 on Build GRCh38
rs756818796 Chr.2: 96253906-96253906 on Build GRCh38
rs200351681 Chr.2: 96253927-96253927 on Build GRCh38
rs746291260 Chr.2: 96253873-96253873 on Build GRCh38
rs776822044 Chr.2: 96253871-96253871 on Build GRCh38
rs761885719 Chr.2: 96253980-96253980 on Build GRCh38
rs994504317 Chr.2: 96254025-96254025 on Build GRCh38
rs373951977 Chr.2: 96253860-96253860 on Build GRCh38
rs778160226 Chr.2: 96254020-96254020 on Build GRCh38
rs777748763 Chr.2: 96254043-96254045 on Build GRCh38
rs536921173 Chr.2: 96253961-96253961 on Build GRCh38
rs3852673 Chr.2: 96253904-96253904 on Build GRCh38
rs121908830 Chr.2: 96254050-96254050 on Build GRCh38

View Full Product Details