Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 99156077-99156350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC23
Entrez Gene ID
64927
Gene Name
tetratricopeptide repeat domain 23
Gene Aliases
HCC-8
UniGene
Hs.513195
Gene Chromosome Location
Chr.15: 99136323-99251226 on Build GRCh38
Transcripts
NM_001288615.1 NM_001288616.1 NR_109947.1 XM_011521927.2 XM_011521929.1 XM_011521930.1 XM_011521931.1 XM_011521932.1 XM_011521933.1 XM_011521934.1 XM_011521935.1 XM_011521936.1 XM_011521937.1 XM_011521939.1 XM_011521940.2 XM_011521941.2 XM_011521943.2 XM_011521944.2 XM_011521945.2 XM_017022509.1 XM_017022510.1 XM_017022511.1 XM_017022512.1 XM_017022513.1 XM_017022514.1 XM_017022515.1 XM_017022516.1 XM_017022517.1 XM_017022518.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3936907 c.1103C>G p.A368G Chr.15: 99156188-99156188 on Build GRCh38
2141608 c.1070A>G p.E357G Chr.15: 99156221-99156221 on Build GRCh38
5659959 c.1115A>T p.H372L Chr.15: 99156176-99156176 on Build GRCh38
5659958 c.1115A>T p.H372L Chr.15: 99156176-99156176 on Build GRCh38
4255172 c.1070A>G p.E357G Chr.15: 99156221-99156221 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs981710569 Chr.15: 99156129-99156129 on Build GRCh38
rs751069510 Chr.15: 99156192-99156192 on Build GRCh38
rs761355684 Chr.15: 99156119-99156119 on Build GRCh38
rs776611799 Chr.15: 99156250-99156250 on Build GRCh38
rs775602522 Chr.15: 99156274-99156275 on Build GRCh38
rs760398305 Chr.15: 99156244-99156244 on Build GRCh38
rs754459224 Chr.15: 99156276-99156276 on Build GRCh38
rs151128482 Chr.15: 99156275-99156275 on Build GRCh38
rs757392304 Chr.15: 99156172-99156172 on Build GRCh38
rs767409478 Chr.15: 99156122-99156122 on Build GRCh38
rs760721072 Chr.15: 99156302-99156303 on Build GRCh38
rs547786597 Chr.15: 99156286-99156286 on Build GRCh38
rs77414166 Chr.15: 99156308-99156308 on Build GRCh38
rs755404400 Chr.15: 99156283-99156283 on Build GRCh38
rs957011890 Chr.15: 99156243-99156243 on Build GRCh38
rs1012942862 Chr.15: 99156324-99156324 on Build GRCh38
rs775161865 Chr.15: 99156100-99156100 on Build GRCh38
rs924313077 Chr.15: 99156137-99156137 on Build GRCh38
rs759387845 Chr.15: 99156251-99156251 on Build GRCh38
rs748766124 Chr.15: 99156114-99156114 on Build GRCh38
rs758420375 Chr.15: 99156144-99156144 on Build GRCh38
rs756487737 Chr.15: 99156267-99156267 on Build GRCh38
rs138499311 Chr.15: 99156196-99156196 on Build GRCh38
rs889079541 Chr.15: 99156175-99156175 on Build GRCh38
rs369825526 Chr.15: 99156287-99156287 on Build GRCh38
rs369463245 Chr.15: 99156116-99156116 on Build GRCh38
rs866559100 Chr.15: 99156234-99156234 on Build GRCh38
rs34584040 Chr.15: 99156145-99156146 on Build GRCh38
rs201596385 Chr.15: 99156147-99156147 on Build GRCh38
rs762119290 Chr.15: 99156258-99156258 on Build GRCh38
rs765736859 Chr.15: 99156187-99156187 on Build GRCh38
rs749224384 Chr.15: 99156165-99156165 on Build GRCh38
rs548711410 Chr.15: 99156113-99156113 on Build GRCh38
rs200643598 Chr.15: 99156123-99156123 on Build GRCh38
rs751877078 Chr.15: 99156165-99156165 on Build GRCh38
rs748821272 Chr.15: 99156212-99156212 on Build GRCh38
rs199700206 Chr.15: 99156118-99156118 on Build GRCh38
rs751571056 Chr.15: 99156252-99156252 on Build GRCh38
rs201912078 Chr.15: 99156245-99156245 on Build GRCh38
rs781658930 Chr.15: 99156173-99156173 on Build GRCh38
rs770849993 Chr.15: 99156258-99156259 on Build GRCh38
rs199623284 Chr.15: 99156176-99156176 on Build GRCh38
rs757937021 Chr.15: 99156164-99156164 on Build GRCh38
rs571528664 Chr.15: 99156217-99156217 on Build GRCh38
rs750820537 Chr.15: 99156265-99156265 on Build GRCh38
rs749702238 Chr.15: 99156316-99156316 on Build GRCh38
rs770780457 Chr.15: 99156298-99156298 on Build GRCh38
rs752782708 Chr.15: 99156136-99156136 on Build GRCh38
rs756736986 Chr.15: 99156231-99156231 on Build GRCh38
rs777351335 Chr.15: 99156291-99156291 on Build GRCh38
rs778079879 Chr.15: 99156218-99156218 on Build GRCh38
rs768266565 Chr.15: 99156213-99156213 on Build GRCh38
rs773024985 Chr.15: 99156232-99156232 on Build GRCh38
rs142969766 Chr.15: 99156226-99156226 on Build GRCh38
rs930691377 Chr.15: 99156160-99156160 on Build GRCh38
rs767805752 Chr.15: 99156260-99156260 on Build GRCh38
rs776380639 Chr.15: 99156299-99156299 on Build GRCh38
rs147497184 Chr.15: 99156185-99156185 on Build GRCh38

View Full Product Details