Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 63174489-63174762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDPCP
Entrez Gene ID
51057
Gene Name
WD repeat containing planar cell polarity effector
Gene Aliases
BBS15, C2orf86, CHDTHP, FRITZ, FRTZ
UniGene
Hs.414952
Gene Chromosome Location
Chr.2: 63120755-63840825 on Build GRCh38
Transcripts
NM_001042692.2 NM_015910.5 NR_122106.1 XM_005264348.3 XM_011532881.2 XM_017004251.1 XM_017004252.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3054880 c.2009A>T p.N670I Chr.2: 63174739-63174739 on Build GRCh38
1022195 c.2060T>C p.L687P Chr.2: 63174688-63174688 on Build GRCh38
259813 c.2002C>T p.P668S Chr.2: 63174746-63174746 on Build GRCh38
3372839 c.1988G>C p.G663A Chr.2: 63174760-63174760 on Build GRCh38
7167804 c.2036G>A p.R679K Chr.2: 63174712-63174712 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749097497 Chr.2: 63174645-63174645 on Build GRCh38
rs975020935 Chr.2: 63174546-63174546 on Build GRCh38
rs371652879 Chr.2: 63174651-63174651 on Build GRCh38
rs954670149 Chr.2: 63174549-63174549 on Build GRCh38
rs759847898 Chr.2: 63174700-63174700 on Build GRCh38
rs552902874 Chr.2: 63174571-63174571 on Build GRCh38
rs934670040 Chr.2: 63174627-63174627 on Build GRCh38
rs777876766 Chr.2: 63174624-63174624 on Build GRCh38
rs757539238 Chr.2: 63174631-63174631 on Build GRCh38
rs758998604 Chr.2: 63174718-63174718 on Build GRCh38
rs766136478 Chr.2: 63174680-63174680 on Build GRCh38
rs755528724 Chr.2: 63174641-63174641 on Build GRCh38
rs765674630 Chr.2: 63174701-63174701 on Build GRCh38
rs372440565 Chr.2: 63174545-63174545 on Build GRCh38
rs924589632 Chr.2: 63174607-63174607 on Build GRCh38
rs781349761 Chr.2: 63174670-63174671 on Build GRCh38
rs771461774 Chr.2: 63174657-63174657 on Build GRCh38
rs760401387 Chr.2: 63174673-63174673 on Build GRCh38
rs748159985 Chr.2: 63174652-63174652 on Build GRCh38
rs571374696 Chr.2: 63174598-63174598 on Build GRCh38
rs755360318 Chr.2: 63174659-63174659 on Build GRCh38
rs61734468 Chr.2: 63174685-63174685 on Build GRCh38
rs772915355 Chr.2: 63174658-63174658 on Build GRCh38
rs778696503 Chr.2: 63174558-63174558 on Build GRCh38
rs779754761 Chr.2: 63174637-63174637 on Build GRCh38
rs768591606 Chr.2: 63174649-63174649 on Build GRCh38
rs753121252 Chr.2: 63174717-63174717 on Build GRCh38
rs915151252 Chr.2: 63174554-63174554 on Build GRCh38
rs376702516 Chr.2: 63174719-63174719 on Build GRCh38
rs538569695 Chr.2: 63174608-63174608 on Build GRCh38

View Full Product Details