Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 44320458-44320683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PREPL
Entrez Gene ID
9581
Gene Name
prolyl endopeptidase-like
Gene Aliases
-
UniGene
Hs.444349
Gene Chromosome Location
Chr.2: 44317607-44361862 on Build GRCh38
Transcripts
NM_001042385.2 NM_001042386.2 NM_001171603.1 NM_001171606.1 NM_001171613.1 NM_001171617.1 NM_006036.4 XM_011533198.1 XM_011533200.1 XM_011533202.1 XM_017005384.1 XM_017005385.1 NM_000341.3

Gene Symbol
SLC3A1
Entrez Gene ID
6519
Gene Name
solute carrier family 3 member 1
Gene Aliases
ATR1, CSNU1, D2H, NBAT, RBAT
UniGene
Hs.112916
Gene Chromosome Location
Chr.2: 44275458-44320824 on Build GRCh38
Transcripts
NM_001042385.2 NM_001042386.2 NM_001171603.1 NM_001171606.1 NM_001171613.1 NM_001171617.1 NM_006036.4 XM_011533198.1 XM_011533200.1 XM_011533202.1 XM_017005384.1 XM_017005385.1 NM_000341.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3185318 c.1956G>A p.T652T Chr.2: 44320537-44320537 on Build GRCh38
3839580 c.2044T>C p.Y682H Chr.2: 44320625-44320625 on Build GRCh38
138298 c.1928G>A p.G643E Chr.2: 44320509-44320509 on Build GRCh38
477441 c.2040A>G p.I680M Chr.2: 44320621-44320621 on Build GRCh38
3185316 c.1881C>T p.A627A Chr.2: 44320462-44320462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779104116 Chr.2: 44320625-44320625 on Build GRCh38
rs148449612 Chr.2: 44320581-44320581 on Build GRCh38
rs747267676 Chr.2: 44320635-44320635 on Build GRCh38
rs761263537 Chr.2: 44320573-44320573 on Build GRCh38
rs771277513 Chr.2: 44320640-44320640 on Build GRCh38
rs769571471 Chr.2: 44320545-44320545 on Build GRCh38
rs777149216 Chr.2: 44320600-44320601 on Build GRCh38
rs924248668 Chr.2: 44320619-44320619 on Build GRCh38
rs780821343 Chr.2: 44320537-44320537 on Build GRCh38
rs767011523 Chr.2: 44320662-44320662 on Build GRCh38
rs749314531 Chr.2: 44320553-44320553 on Build GRCh38
rs962167700 Chr.2: 44320583-44320583 on Build GRCh38
rs751203733 Chr.2: 44320535-44320535 on Build GRCh38
rs772132458 Chr.2: 44320632-44320632 on Build GRCh38
rs121912695 Chr.2: 44320536-44320536 on Build GRCh38
rs764488098 Chr.2: 44320580-44320581 on Build GRCh38
rs563709928 Chr.2: 44320555-44320555 on Build GRCh38
rs376083735 Chr.2: 44320488-44320488 on Build GRCh38
rs149749634 Chr.2: 44320534-44320534 on Build GRCh38
rs371136651 Chr.2: 44320594-44320594 on Build GRCh38
rs955228283 Chr.2: 44320513-44320513 on Build GRCh38
rs896210706 Chr.2: 44320491-44320491 on Build GRCh38
rs376897765 Chr.2: 44320618-44320618 on Build GRCh38
rs748433525 Chr.2: 44320630-44320630 on Build GRCh38
rs763629706 Chr.2: 44320645-44320645 on Build GRCh38
rs766179607 Chr.2: 44320529-44320529 on Build GRCh38
rs756823144 Chr.2: 44320598-44320598 on Build GRCh38
rs761541250 Chr.2: 44320661-44320661 on Build GRCh38
rs376270408 Chr.2: 44320603-44320603 on Build GRCh38
rs773635956 Chr.2: 44320633-44320633 on Build GRCh38
rs777174361 Chr.2: 44320643-44320643 on Build GRCh38
rs199949010 Chr.2: 44320560-44320560 on Build GRCh38
rs121912693 Chr.2: 44320614-44320614 on Build GRCh38
rs762511951 Chr.2: 44320593-44320593 on Build GRCh38
rs975911420 Chr.2: 44320586-44320586 on Build GRCh38
rs267599386 Chr.2: 44320502-44320502 on Build GRCh38
rs373698143 Chr.2: 44320563-44320563 on Build GRCh38
rs751254985 Chr.2: 44320595-44320595 on Build GRCh38
rs542758042 Chr.2: 44320623-44320623 on Build GRCh38
rs750128585 Chr.2: 44320608-44320608 on Build GRCh38
rs35046592 Chr.2: 44320569-44320569 on Build GRCh38
rs756083951 Chr.2: 44320613-44320613 on Build GRCh38
rs368784606 Chr.2: 44320588-44320588 on Build GRCh38
rs377448239 Chr.2: 44320572-44320572 on Build GRCh38
rs370262167 Chr.2: 44320554-44320554 on Build GRCh38
rs745852156 Chr.2: 44320543-44320543 on Build GRCh38
rs558927581 Chr.2: 44320485-44320485 on Build GRCh38
rs767168885 Chr.2: 44320577-44320577 on Build GRCh38
rs780297364 Chr.2: 44320552-44320552 on Build GRCh38
rs773806097 Chr.2: 44320657-44320657 on Build GRCh38

View Full Product Details