Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68303821-68304076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC7A6OS
Entrez Gene ID
84138
Gene Name
solute carrier family 7 member 6 opposite strand
Gene Aliases
-
UniGene
Hs.729520
Gene Chromosome Location
Chr.16: 68300615-68310965 on Build GRCh38
Transcripts
NM_032178.2 XM_011523372.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5929685 c.672G>A p.W224* Chr.16: 68304032-68304032 on Build GRCh38
972730 c.649G>A p.V217M Chr.16: 68304055-68304055 on Build GRCh38
1178914 c.645C>T p.L215L Chr.16: 68304059-68304059 on Build GRCh38
435575 c.662G>T p.S221I Chr.16: 68304042-68304042 on Build GRCh38
5929686 c.671G>A p.W224* Chr.16: 68304033-68304033 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1015148866 Chr.16: 68303943-68303943 on Build GRCh38
rs976210250 Chr.16: 68303955-68303955 on Build GRCh38
rs374727375 Chr.16: 68303960-68303960 on Build GRCh38
rs962617825 Chr.16: 68303947-68303947 on Build GRCh38
rs372590458 Chr.16: 68303993-68303993 on Build GRCh38
rs772848364 Chr.16: 68304013-68304023 on Build GRCh38
rs549790098 Chr.16: 68303882-68303882 on Build GRCh38
rs193289010 Chr.16: 68303939-68303939 on Build GRCh38
rs6499167 Chr.16: 68303895-68303895 on Build GRCh38
rs558919716 Chr.16: 68303958-68303958 on Build GRCh38
rs7200884 Chr.16: 68304004-68304004 on Build GRCh38
rs144208197 Chr.16: 68304003-68304003 on Build GRCh38
rs535699453 Chr.16: 68303931-68303931 on Build GRCh38
rs372147827 Chr.16: 68304011-68304011 on Build GRCh38
rs1046371804 Chr.16: 68304024-68304024 on Build GRCh38
rs755124077 Chr.16: 68303945-68303945 on Build GRCh38
rs202170729 Chr.16: 68304034-68304034 on Build GRCh38
rs539241288 Chr.16: 68303967-68303967 on Build GRCh38
rs781199487 Chr.16: 68303997-68303997 on Build GRCh38
rs11548855 Chr.16: 68304045-68304045 on Build GRCh38
rs751483213 Chr.16: 68303981-68303981 on Build GRCh38
rs543519497 Chr.16: 68304006-68304006 on Build GRCh38
rs759492044 Chr.16: 68303978-68303978 on Build GRCh38
rs779491490 Chr.16: 68304029-68304029 on Build GRCh38
rs766485012 Chr.16: 68303975-68303975 on Build GRCh38
rs771210829 Chr.16: 68304050-68304050 on Build GRCh38
rs143239221 Chr.16: 68304035-68304035 on Build GRCh38
rs754274503 Chr.16: 68303941-68303941 on Build GRCh38
rs768005272 Chr.16: 68304017-68304017 on Build GRCh38
rs761308489 Chr.16: 68303861-68303861 on Build GRCh38
rs753987929 Chr.16: 68303976-68303976 on Build GRCh38
rs764112242 Chr.16: 68303979-68303979 on Build GRCh38
rs531477472 Chr.16: 68303871-68303871 on Build GRCh38

View Full Product Details