Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46832760-46833033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRSS42
Entrez Gene ID
339906
Gene Name
protease, serine 42
Gene Aliases
TESSP2
UniGene
Hs.585127
Gene Chromosome Location
Chr.3: 46830404-46834095 on Build GRCh38
Transcripts
NM_182702.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
84798 c.602G>T p.W201L Chr.3: 46832976-46832976 on Build GRCh38
6097943 c.596G>T p.R199M Chr.3: 46832982-46832982 on Build GRCh38
1423455 c.621A>C p.K207N Chr.3: 46832957-46832957 on Build GRCh38
6884218 c.603G>A p.W201* Chr.3: 46832975-46832975 on Build GRCh38
6231804 c.586G>T p.G196C Chr.3: 46832992-46832992 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs535420967 Chr.3: 46832815-46832815 on Build GRCh38
rs1043171674 Chr.3: 46832863-46832863 on Build GRCh38
rs771539610 Chr.3: 46832839-46832839 on Build GRCh38
rs549146042 Chr.3: 46832851-46832851 on Build GRCh38
rs1035450567 Chr.3: 46832928-46832928 on Build GRCh38
rs748451747 Chr.3: 46832988-46832988 on Build GRCh38
rs968330431 Chr.3: 46832959-46832959 on Build GRCh38
rs374513051 Chr.3: 46833009-46833009 on Build GRCh38
rs757136432 Chr.3: 46832968-46832968 on Build GRCh38
rs755607767 Chr.3: 46832952-46832952 on Build GRCh38
rs763489988 Chr.3: 46832938-46832938 on Build GRCh38
rs768718703 Chr.3: 46832914-46832914 on Build GRCh38
rs770858562 Chr.3: 46832897-46832897 on Build GRCh38
rs745373393 Chr.3: 46832797-46832797 on Build GRCh38
rs995447671 Chr.3: 46832858-46832858 on Build GRCh38
rs186113871 Chr.3: 46832908-46832908 on Build GRCh38
rs890496658 Chr.3: 46832814-46832814 on Build GRCh38
rs751824490 Chr.3: 46832946-46832946 on Build GRCh38
rs200671288 Chr.3: 46832993-46832993 on Build GRCh38
rs904596849 Chr.3: 46832881-46832881 on Build GRCh38
rs745616564 Chr.3: 46832975-46832975 on Build GRCh38
rs370252644 Chr.3: 46832950-46832950 on Build GRCh38
rs776949063 Chr.3: 46832916-46832916 on Build GRCh38
rs1026881886 Chr.3: 46832869-46832869 on Build GRCh38
rs528661411 Chr.3: 46832792-46832792 on Build GRCh38
rs777197028 Chr.3: 46832960-46832960 on Build GRCh38
rs748810036 Chr.3: 46832961-46832961 on Build GRCh38
rs776700321 Chr.3: 46832986-46832986 on Build GRCh38
rs771948681 Chr.3: 46832979-46832979 on Build GRCh38
rs200786285 Chr.3: 46832937-46832937 on Build GRCh38
rs548119689 Chr.3: 46832812-46832812 on Build GRCh38
rs763150366 Chr.3: 46833013-46833013 on Build GRCh38
rs899151832 Chr.3: 46832852-46832852 on Build GRCh38
rs993120795 Chr.3: 46832794-46832794 on Build GRCh38
rs951208274 Chr.3: 46832896-46832896 on Build GRCh38
rs767048114 Chr.3: 46832947-46832947 on Build GRCh38
rs765461661 Chr.3: 46832936-46832936 on Build GRCh38
rs371004657 Chr.3: 46832791-46832791 on Build GRCh38
rs767010325 Chr.3: 46832939-46832939 on Build GRCh38
rs570738558 Chr.3: 46832932-46832932 on Build GRCh38
rs778939590 Chr.3: 46832969-46832969 on Build GRCh38

View Full Product Details