Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 100037589-100037848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UHRF1BP1L
Entrez Gene ID
23074
Gene Name
UHRF1 binding protein 1 like
Gene Aliases
SHIP164
UniGene
Hs.572154
Gene Chromosome Location
Chr.12: 100026251-100142864 on Build GRCh38
Transcripts
NM_015054.1 XM_005268738.2 XM_005268739.3 XM_011538054.2 XM_017019047.1 XM_017019048.1 XM_017019050.1 XM_017019051.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4652124 c.4364T>A p.L1455H Chr.12: 100037667-100037667 on Build GRCh38
4967008 c.4315A>C p.K1439Q Chr.12: 100037716-100037716 on Build GRCh38
7137711 c.4348G>C p.E1450Q Chr.12: 100037683-100037683 on Build GRCh38
690832 c.4331T>C p.L1444P Chr.12: 100037700-100037700 on Build GRCh38
6490080 c.4276C>G p.L1426V Chr.12: 100037755-100037755 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111388230 Chr.12: 100037772-100037772 on Build GRCh38
rs752411777 Chr.12: 100037665-100037667 on Build GRCh38
rs746240661 Chr.12: 100037748-100037748 on Build GRCh38
rs12310953 Chr.12: 100037773-100037773 on Build GRCh38
rs148413208 Chr.12: 100037752-100037752 on Build GRCh38
rs903617318 Chr.12: 100037755-100037755 on Build GRCh38
rs750364318 Chr.12: 100037621-100037621 on Build GRCh38
rs759391654 Chr.12: 100037785-100037785 on Build GRCh38
rs370385647 Chr.12: 100037616-100037616 on Build GRCh38
rs772336254 Chr.12: 100037762-100037762 on Build GRCh38
rs766306081 Chr.12: 100037777-100037777 on Build GRCh38
rs771447956 Chr.12: 100037734-100037734 on Build GRCh38
rs764101936 Chr.12: 100037620-100037620 on Build GRCh38
rs368712199 Chr.12: 100037659-100037659 on Build GRCh38
rs761015837 Chr.12: 100037772-100037773 on Build GRCh38
rs779187131 Chr.12: 100037737-100037737 on Build GRCh38
rs754624334 Chr.12: 100037644-100037644 on Build GRCh38
rs777382260 Chr.12: 100037670-100037670 on Build GRCh38
rs747166594 Chr.12: 100037771-100037771 on Build GRCh38
rs774074996 Chr.12: 100037779-100037779 on Build GRCh38
rs964294011 Chr.12: 100037819-100037819 on Build GRCh38
rs144439125 Chr.12: 100037647-100037647 on Build GRCh38
rs765394183 Chr.12: 100037639-100037639 on Build GRCh38
rs749782535 Chr.12: 100037722-100037722 on Build GRCh38
rs984284228 Chr.12: 100037815-100037815 on Build GRCh38
rs148072766 Chr.12: 100037817-100037817 on Build GRCh38
rs780980450 Chr.12: 100037645-100037645 on Build GRCh38
rs372236461 Chr.12: 100037667-100037667 on Build GRCh38
rs762904429 Chr.12: 100037624-100037624 on Build GRCh38
rs766106475 Chr.12: 100037632-100037632 on Build GRCh38
rs143232397 Chr.12: 100037765-100037765 on Build GRCh38
rs373807346 Chr.12: 100037657-100037657 on Build GRCh38
rs764218431 Chr.12: 100037788-100037788 on Build GRCh38
rs751343416 Chr.12: 100037642-100037642 on Build GRCh38

View Full Product Details