Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 134191712-134191957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RYK
Entrez Gene ID
6259
Gene Name
receptor-like tyrosine kinase
Gene Aliases
D3S3195, JTK5, JTK5A, RYK1
UniGene
Hs.654562
Gene Chromosome Location
Chr.3: 134157133-134250834 on Build GRCh38
Transcripts
NM_001005861.2 NM_002958.3 XM_017007016.1

SNP Detail

SNP ID Chromosome Location
rs372101827 Chr.3: 134191923-134191923 on Build GRCh38
rs912050878 Chr.3: 134191783-134191783 on Build GRCh38
rs778701959 Chr.3: 134191916-134191916 on Build GRCh38
rs370703140 Chr.3: 134191858-134191858 on Build GRCh38
rs995457362 Chr.3: 134191750-134191750 on Build GRCh38
rs147089787 Chr.3: 134191829-134191829 on Build GRCh38
rs748945264 Chr.3: 134191906-134191906 on Build GRCh38
rs537570512 Chr.3: 134191797-134191797 on Build GRCh38
rs1042591532 Chr.3: 134191769-134191769 on Build GRCh38
rs577277523 Chr.3: 134191839-134191839 on Build GRCh38
rs780654400 Chr.3: 134191878-134191878 on Build GRCh38
rs764210355 Chr.3: 134191845-134191845 on Build GRCh38
rs368346060 Chr.3: 134191880-134191880 on Build GRCh38
rs772141154 Chr.3: 134191837-134191837 on Build GRCh38
rs953929849 Chr.3: 134191789-134191789 on Build GRCh38
rs777289155 Chr.3: 134191889-134191889 on Build GRCh38
rs774206383 Chr.3: 134191828-134191828 on Build GRCh38
rs554508310 Chr.3: 134191787-134191787 on Build GRCh38
rs762204556 Chr.3: 134191862-134191862 on Build GRCh38
rs770776836 Chr.3: 134191813-134191813 on Build GRCh38
rs769297012 Chr.3: 134191802-134191802 on Build GRCh38
rs568525826 Chr.3: 134191792-134191792 on Build GRCh38
rs770656029 Chr.3: 134191913-134191913 on Build GRCh38
rs915495933 Chr.3: 134191747-134191747 on Build GRCh38
rs937537260 Chr.3: 134191900-134191900 on Build GRCh38
rs775554119 Chr.3: 134191927-134191927 on Build GRCh38
rs759001453 Chr.3: 134191870-134191870 on Build GRCh38
rs886371658 Chr.3: 134191856-134191856 on Build GRCh38
rs61732795 Chr.3: 134191925-134191925 on Build GRCh38
rs945584737 Chr.3: 134191751-134191751 on Build GRCh38
rs985687139 Chr.3: 134191790-134191790 on Build GRCh38
rs912662449 Chr.3: 134191795-134191795 on Build GRCh38
rs765713202 Chr.3: 134191865-134191865 on Build GRCh38
rs772963317 Chr.3: 134191806-134191806 on Build GRCh38
rs375133082 Chr.3: 134191877-134191877 on Build GRCh38
rs755710425 Chr.3: 134191888-134191888 on Build GRCh38
rs55965934 Chr.3: 134191843-134191843 on Build GRCh38

View Full Product Details