Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 13719854-13720099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAPPC2
Entrez Gene ID
6399
Gene Name
trafficking protein particle complex 2
Gene Aliases
MIP2A, SEDL, SEDT, TRAPPC2P1, TRS20, ZNF547L, hYP38334
UniGene
Hs.592238
Gene Chromosome Location
Chr.X: 13712242-13734635 on Build GRCh38
Transcripts
NM_001011658.3 NM_001128835.2 NM_014563.5 XM_011545565.1 XM_011545566.2

SNP Detail

SNP ID Chromosome Location
rs768120589 Chr.X: 13719926-13719926 on Build GRCh38
rs768424215 Chr.X: 13720015-13720015 on Build GRCh38
rs777366779 Chr.X: 13719892-13719892 on Build GRCh38
rs1040626791 Chr.X: 13719900-13719900 on Build GRCh38
rs773141571 Chr.X: 13719891-13719892 on Build GRCh38
rs587776748 Chr.X: 13719910-13719911 on Build GRCh38
rs770205303 Chr.X: 13720021-13720021 on Build GRCh38
rs780179384 Chr.X: 13719916-13719916 on Build GRCh38
rs887906785 Chr.X: 13720053-13720053 on Build GRCh38
rs931966637 Chr.X: 13719919-13719919 on Build GRCh38
rs753831612 Chr.X: 13719904-13719904 on Build GRCh38
rs770233317 Chr.X: 13719980-13719980 on Build GRCh38
rs921928773 Chr.X: 13719905-13719905 on Build GRCh38
rs867735712 Chr.X: 13719894-13719894 on Build GRCh38
rs757131748 Chr.X: 13719909-13719909 on Build GRCh38
rs987057675 Chr.X: 13720036-13720036 on Build GRCh38
rs768670689 Chr.X: 13719966-13719966 on Build GRCh38
rs747064126 Chr.X: 13719947-13719947 on Build GRCh38
rs781420575 Chr.X: 13719985-13719985 on Build GRCh38

View Full Product Details