Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57888616-57888871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF814
Entrez Gene ID
730051
Gene Name
zinc finger protein 814
Gene Aliases
-
UniGene
Hs.634143
Gene Chromosome Location
Chr.19: 57869379-57889074 on Build GRCh38
Transcripts
NM_001144989.1 XM_017027197.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6585443 c.27C>G p.L9L Chr.19: 57888776-57888776 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777212502 Chr.19: 57888847-57888847 on Build GRCh38
rs559059271 Chr.19: 57888679-57888679 on Build GRCh38
rs982812722 Chr.19: 57888818-57888818 on Build GRCh38
rs771906185 Chr.19: 57888829-57888829 on Build GRCh38
rs751783540 Chr.19: 57888665-57888665 on Build GRCh38
rs547364165 Chr.19: 57888838-57888838 on Build GRCh38
rs74374013 Chr.19: 57888830-57888830 on Build GRCh38
rs759638827 Chr.19: 57888727-57888727 on Build GRCh38
rs376801406 Chr.19: 57888794-57888794 on Build GRCh38
rs1007613349 Chr.19: 57888746-57888746 on Build GRCh38
rs982864956 Chr.19: 57888638-57888638 on Build GRCh38
rs950275714 Chr.19: 57888833-57888833 on Build GRCh38
rs545157975 Chr.19: 57888795-57888795 on Build GRCh38
rs3948443 Chr.19: 57888824-57888824 on Build GRCh38
rs375550431 Chr.19: 57888750-57888750 on Build GRCh38
rs1054641297 Chr.19: 57888736-57888736 on Build GRCh38
rs530928975 Chr.19: 57888825-57888825 on Build GRCh38
rs934837989 Chr.19: 57888843-57888843 on Build GRCh38
rs565344196 Chr.19: 57888820-57888820 on Build GRCh38
rs765633769 Chr.19: 57888852-57888852 on Build GRCh38
rs35114097 Chr.19: 57888776-57888776 on Build GRCh38
rs759755372 Chr.19: 57888849-57888849 on Build GRCh38
rs747142038 Chr.19: 57888785-57888785 on Build GRCh38
rs909873148 Chr.19: 57888654-57888654 on Build GRCh38
rs755396820 Chr.19: 57888734-57888734 on Build GRCh38
rs530408079 Chr.19: 57888836-57888836 on Build GRCh38
rs550901638 Chr.19: 57888828-57888828 on Build GRCh38
rs998597126 Chr.19: 57888733-57888733 on Build GRCh38
rs767037360 Chr.19: 57888723-57888723 on Build GRCh38
rs142144396 Chr.19: 57888791-57888791 on Build GRCh38
rs997429472 Chr.19: 57888786-57888786 on Build GRCh38
rs924759664 Chr.19: 57888839-57888839 on Build GRCh38
rs34907069 Chr.19: 57888780-57888781 on Build GRCh38
rs767532928 Chr.19: 57888740-57888740 on Build GRCh38
rs777817904 Chr.19: 57888773-57888773 on Build GRCh38
rs138986175 Chr.19: 57888690-57888690 on Build GRCh38
rs752851883 Chr.19: 57888813-57888813 on Build GRCh38
rs1046046082 Chr.19: 57888837-57888837 on Build GRCh38
rs892784186 Chr.19: 57888741-57888741 on Build GRCh38
rs75177271 Chr.19: 57888731-57888731 on Build GRCh38
rs778360839 Chr.19: 57888827-57888827 on Build GRCh38
rs965894988 Chr.19: 57888777-57888777 on Build GRCh38

View Full Product Details