Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 42224432-42224667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF526
Entrez Gene ID
116115
Gene Name
zinc finger protein 526
Gene Aliases
-
UniGene
Hs.137282
Gene Chromosome Location
Chr.19: 42220271-42227053 on Build GRCh38
Transcripts
NM_001314033.1 NM_133444.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6830956 c.52G>T p.G18W Chr.19: 42224455-42224455 on Build GRCh38
5936563 c.148C>T p.L50F Chr.19: 42224551-42224551 on Build GRCh38
6241445 c.241C>A p.L81I Chr.19: 42224644-42224644 on Build GRCh38
3797136 c.253G>A p.E85K Chr.19: 42224656-42224656 on Build GRCh38
4131926 c.255G>A p.E85E Chr.19: 42224658-42224658 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778028127 Chr.19: 42224554-42224554 on Build GRCh38
rs1057495825 Chr.19: 42224551-42224551 on Build GRCh38
rs980900134 Chr.19: 42224639-42224639 on Build GRCh38
rs149913848 Chr.19: 42224484-42224484 on Build GRCh38
rs993134546 Chr.19: 42224526-42224526 on Build GRCh38
rs772553789 Chr.19: 42224594-42224594 on Build GRCh38
rs749361401 Chr.19: 42224571-42224571 on Build GRCh38
rs769360796 Chr.19: 42224608-42224608 on Build GRCh38
rs969088386 Chr.19: 42224572-42224572 on Build GRCh38
rs904672368 Chr.19: 42224456-42224456 on Build GRCh38
rs757883823 Chr.19: 42224497-42224497 on Build GRCh38
rs746308322 Chr.19: 42224591-42224591 on Build GRCh38
rs201301381 Chr.19: 42224483-42224483 on Build GRCh38
rs146838645 Chr.19: 42224506-42224506 on Build GRCh38
rs770996097 Chr.19: 42224582-42224582 on Build GRCh38
rs759917688 Chr.19: 42224465-42224465 on Build GRCh38
rs747946795 Chr.19: 42224518-42224518 on Build GRCh38
rs762380036 Chr.19: 42224628-42224628 on Build GRCh38
rs774853837 Chr.19: 42224584-42224584 on Build GRCh38
rs775827458 Chr.19: 42224481-42224481 on Build GRCh38
rs766247400 Chr.19: 42224502-42224502 on Build GRCh38
rs775882267 Chr.19: 42224603-42224603 on Build GRCh38
rs774800827 Chr.19: 42224459-42224459 on Build GRCh38
rs771384051 Chr.19: 42224453-42224453 on Build GRCh38
rs754821112 Chr.19: 42224508-42224508 on Build GRCh38
rs749939180 Chr.19: 42224493-42224493 on Build GRCh38
rs948386196 Chr.19: 42224616-42224616 on Build GRCh38
rs761067223 Chr.19: 42224607-42224607 on Build GRCh38
rs139088396 Chr.19: 42224479-42224479 on Build GRCh38
rs1042750189 Chr.19: 42224640-42224640 on Build GRCh38
rs756343377 Chr.19: 42224538-42224538 on Build GRCh38

View Full Product Details