Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49184902-49185175 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUBA1A
Entrez Gene ID
7846
Gene Name
tubulin alpha 1a
Gene Aliases
B-ALPHA-1, LIS3, TUBA3
UniGene
Hs.654422
Gene Chromosome Location
Chr.12: 49184795-49189324 on Build GRCh38
Transcripts
NM_001270399.1 NM_001270400.1 NM_006009.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1195822 c.1205G>A p.R402H Chr.12: 49185161-49185161 on Build GRCh38
5035540 c.1215T>G p.V405V Chr.12: 49185151-49185151 on Build GRCh38
5375291 c.1348G>A p.E450K Chr.12: 49185018-49185018 on Build GRCh38
4920884 c.1274T>C p.M425T Chr.12: 49185092-49185092 on Build GRCh38
7287864 c.1248T>C p.G416G Chr.12: 49185118-49185118 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs960498970 Chr.12: 49184940-49184940 on Build GRCh38
rs185938303 Chr.12: 49184984-49184984 on Build GRCh38
rs772809398 Chr.12: 49185136-49185136 on Build GRCh38
rs137853047 Chr.12: 49185110-49185110 on Build GRCh38
rs753757160 Chr.12: 49184988-49184988 on Build GRCh38
rs781483247 Chr.12: 49185009-49185009 on Build GRCh38
rs753719501 Chr.12: 49185142-49185142 on Build GRCh38
rs11558070 Chr.12: 49185103-49185103 on Build GRCh38
rs371718931 Chr.12: 49185001-49185001 on Build GRCh38
rs777929464 Chr.12: 49184994-49184994 on Build GRCh38
rs137853050 Chr.12: 49185101-49185101 on Build GRCh38
rs749746325 Chr.12: 49185048-49185048 on Build GRCh38
rs775885354 Chr.12: 49185037-49185037 on Build GRCh38
rs200873943 Chr.12: 49185019-49185019 on Build GRCh38
rs797045005 Chr.12: 49185140-49185140 on Build GRCh38
rs746276826 Chr.12: 49185031-49185031 on Build GRCh38
rs137853049 Chr.12: 49185102-49185102 on Build GRCh38
rs747080229 Chr.12: 49185000-49185000 on Build GRCh38
rs865953916 Chr.12: 49185109-49185109 on Build GRCh38
rs773920941 Chr.12: 49185086-49185086 on Build GRCh38
rs766255841 Chr.12: 49184987-49184987 on Build GRCh38
rs11558069 Chr.12: 49185046-49185046 on Build GRCh38
rs1062437 Chr.12: 49184997-49184997 on Build GRCh38
rs11558065 Chr.12: 49185108-49185108 on Build GRCh38
rs201884190 Chr.12: 49185112-49185112 on Build GRCh38
rs1065730 Chr.12: 49185027-49185027 on Build GRCh38
rs587784484 Chr.12: 49185092-49185092 on Build GRCh38
rs768060474 Chr.12: 49185067-49185067 on Build GRCh38
rs770156445 Chr.12: 49185035-49185035 on Build GRCh38
rs761301881 Chr.12: 49185100-49185100 on Build GRCh38
rs758417587 Chr.12: 49184991-49184991 on Build GRCh38
rs768839661 Chr.12: 49185006-49185006 on Build GRCh38
rs766135902 Chr.12: 49185139-49185139 on Build GRCh38
rs3206694 Chr.12: 49185118-49185118 on Build GRCh38

View Full Product Details