Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 22813006-22813239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC7A7
Entrez Gene ID
9056
Gene Name
solute carrier family 7 member 7
Gene Aliases
LAT3, LPI, MOP-2, Y+LAT1, y+LAT-1
UniGene
Hs.513147
Gene Chromosome Location
Chr.14: 22773222-22819811 on Build GRCh38
Transcripts
NM_001126105.2 NM_001126106.2 NR_040448.1 XM_006720302.1 XM_011537298.2 XM_011537299.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3495028 c.338G>A p.G113E Chr.14: 22813061-22813061 on Build GRCh38
7191091 c.245T>C p.F82S Chr.14: 22813154-22813154 on Build GRCh38
5811546 c.336A>T p.G112G Chr.14: 22813063-22813063 on Build GRCh38
374162 c.302G>T p.G101V Chr.14: 22813097-22813097 on Build GRCh38
2030540 c.236G>A p.G79E Chr.14: 22813163-22813163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755890052 Chr.14: 22813157-22813157 on Build GRCh38
rs146077876 Chr.14: 22813150-22813150 on Build GRCh38
rs967380531 Chr.14: 22813144-22813144 on Build GRCh38
rs773372874 Chr.14: 22813033-22813033 on Build GRCh38
rs778234328 Chr.14: 22813179-22813179 on Build GRCh38
rs748090602 Chr.14: 22813050-22813050 on Build GRCh38
rs775279224 Chr.14: 22813206-22813206 on Build GRCh38
rs928175691 Chr.14: 22813036-22813036 on Build GRCh38
rs371860097 Chr.14: 22813158-22813158 on Build GRCh38
rs759068554 Chr.14: 22813074-22813074 on Build GRCh38
rs752553654 Chr.14: 22813154-22813154 on Build GRCh38
rs769650911 Chr.14: 22813053-22813053 on Build GRCh38
rs745545300 Chr.14: 22813185-22813185 on Build GRCh38
rs386833814 Chr.14: 22813028-22813028 on Build GRCh38
rs386833812 Chr.14: 22813181-22813184 on Build GRCh38
rs764769171 Chr.14: 22813117-22813117 on Build GRCh38
rs139270936 Chr.14: 22813060-22813060 on Build GRCh38
rs139776370 Chr.14: 22813165-22813165 on Build GRCh38
rs770321396 Chr.14: 22813066-22813066 on Build GRCh38
rs1028584279 Chr.14: 22813070-22813070 on Build GRCh38
rs777682875 Chr.14: 22813057-22813057 on Build GRCh38
rs772506118 Chr.14: 22813081-22813081 on Build GRCh38
rs368317123 Chr.14: 22813149-22813149 on Build GRCh38
rs879187534 Chr.14: 22813092-22813092 on Build GRCh38
rs775943127 Chr.14: 22813086-22813086 on Build GRCh38
rs761270703 Chr.14: 22813102-22813102 on Build GRCh38
rs375791337 Chr.14: 22813130-22813130 on Build GRCh38
rs916075432 Chr.14: 22813211-22813211 on Build GRCh38
rs386833813 Chr.14: 22813144-22813145 on Build GRCh38
rs11568438 Chr.14: 22813127-22813127 on Build GRCh38
rs769986701 Chr.14: 22813203-22813203 on Build GRCh38
rs577167267 Chr.14: 22813071-22813071 on Build GRCh38
rs771561758 Chr.14: 22813200-22813200 on Build GRCh38
rs754387668 Chr.14: 22813126-22813126 on Build GRCh38
rs779253748 Chr.14: 22813041-22813041 on Build GRCh38
rs149810006 Chr.14: 22813171-22813171 on Build GRCh38
rs750589845 Chr.14: 22813047-22813047 on Build GRCh38

View Full Product Details