Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 119755390-119755629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMGCS2
Entrez Gene ID
3158
Gene Name
3-hydroxy-3-methylglutaryl-CoA synthase 2
Gene Aliases
-
UniGene
Hs.59889
Gene Chromosome Location
Chr.1: 119747996-119769125 on Build GRCh38
Transcripts
NM_001166107.1 NM_005518.3 XM_011541313.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6057791 c.1081C>G p.Q361E Chr.1: 119755533-119755533 on Build GRCh38
2179899 c.1176G>A p.S392S Chr.1: 119755438-119755438 on Build GRCh38
4140778 c.1173C>G p.A391A Chr.1: 119755441-119755441 on Build GRCh38
6120846 c.1135_1136GG>TT p.G379L Chr.1: 119755478-119755479 on Build GRCh38
6057792 c.1180C>G p.L394V Chr.1: 119755434-119755434 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111313628 Chr.1: 119755599-119755599 on Build GRCh38
rs780316550 Chr.1: 119755570-119755570 on Build GRCh38
rs774953281 Chr.1: 119755504-119755504 on Build GRCh38
rs771289268 Chr.1: 119755472-119755472 on Build GRCh38
rs772715543 Chr.1: 119755501-119755501 on Build GRCh38
rs375873510 Chr.1: 119755609-119755609 on Build GRCh38
rs777182302 Chr.1: 119755597-119755597 on Build GRCh38
rs587690380 Chr.1: 119755519-119755519 on Build GRCh38
rs764568941 Chr.1: 119755486-119755486 on Build GRCh38
rs766003583 Chr.1: 119755503-119755503 on Build GRCh38
rs746145498 Chr.1: 119755560-119755560 on Build GRCh38
rs781186067 Chr.1: 119755451-119755451 on Build GRCh38
rs769416687 Chr.1: 119755420-119755420 on Build GRCh38
rs772359110 Chr.1: 119755487-119755487 on Build GRCh38
rs746227549 Chr.1: 119755482-119755482 on Build GRCh38
rs761407388 Chr.1: 119755419-119755419 on Build GRCh38
rs747485297 Chr.1: 119755576-119755576 on Build GRCh38
rs764706394 Chr.1: 119755426-119755426 on Build GRCh38
rs762446729 Chr.1: 119755601-119755601 on Build GRCh38
rs1025175254 Chr.1: 119755568-119755568 on Build GRCh38
rs757475645 Chr.1: 119755545-119755545 on Build GRCh38
rs777939704 Chr.1: 119755528-119755528 on Build GRCh38
rs991904954 Chr.1: 119755531-119755531 on Build GRCh38
rs753951704 Chr.1: 119755534-119755534 on Build GRCh38
rs4716 Chr.1: 119755461-119755461 on Build GRCh38
rs752639075 Chr.1: 119755522-119755522 on Build GRCh38
rs756169207 Chr.1: 119755453-119755453 on Build GRCh38
rs966875458 Chr.1: 119755571-119755571 on Build GRCh38
rs769058291 Chr.1: 119755585-119755585 on Build GRCh38
rs144329134 Chr.1: 119755427-119755427 on Build GRCh38
rs767236647 Chr.1: 119755521-119755521 on Build GRCh38
rs779242610 Chr.1: 119755557-119755557 on Build GRCh38
rs761359337 Chr.1: 119755412-119755412 on Build GRCh38
rs762627608 Chr.1: 119755428-119755428 on Build GRCh38
rs769310958 Chr.1: 119755495-119755495 on Build GRCh38
rs151328418 Chr.1: 119755490-119755490 on Build GRCh38
rs762536796 Chr.1: 119755502-119755502 on Build GRCh38
rs776046708 Chr.1: 119755489-119755489 on Build GRCh38
rs752626288 Chr.1: 119755452-119755452 on Build GRCh38
rs751371132 Chr.1: 119755446-119755446 on Build GRCh38
rs762443891 Chr.1: 119755525-119755525 on Build GRCh38
rs777744720 Chr.1: 119755457-119755457 on Build GRCh38
rs779332852 Chr.1: 119755475-119755475 on Build GRCh38
rs754826235 Chr.1: 119755447-119755447 on Build GRCh38
rs766097440 Chr.1: 119755439-119755439 on Build GRCh38
rs756272484 Chr.1: 119755482-119755483 on Build GRCh38
rs759352736 Chr.1: 119755520-119755520 on Build GRCh38
rs772437190 Chr.1: 119755561-119755561 on Build GRCh38
rs267597953 Chr.1: 119755429-119755429 on Build GRCh38

View Full Product Details