Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240891605-240891878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf54
Entrez Gene ID
79919
Gene Name
chromosome 2 open reading frame 54
Gene Aliases
-
UniGene
Hs.193745
Gene Chromosome Location
Chr.2: 240886048-240896889 on Build GRCh38
Transcripts
NM_001085437.2 NM_001282921.1 NM_024861.3 XM_011511877.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3910019 c.150C>T p.F50F Chr.2: 240891681-240891681 on Build GRCh38
4820504 c.92G>C p.R31T Chr.2: 240891739-240891739 on Build GRCh38
4092934 c.99G>A p.E33E Chr.2: 240891732-240891732 on Build GRCh38
4903913 c.222C>T p.F74F Chr.2: 240891609-240891609 on Build GRCh38
4863627 c.148T>G p.F50V Chr.2: 240891683-240891683 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201722293 Chr.2: 240891644-240891644 on Build GRCh38
rs571841660 Chr.2: 240891783-240891783 on Build GRCh38
rs760294932 Chr.2: 240891633-240891633 on Build GRCh38
rs773413880 Chr.2: 240891785-240891785 on Build GRCh38
rs769392717 Chr.2: 240891855-240891855 on Build GRCh38
rs764432925 Chr.2: 240891645-240891645 on Build GRCh38
rs757359760 Chr.2: 240891649-240891649 on Build GRCh38
rs745313572 Chr.2: 240891849-240891849 on Build GRCh38
rs377523321 Chr.2: 240891646-240891646 on Build GRCh38
rs199581838 Chr.2: 240891758-240891758 on Build GRCh38
rs374993520 Chr.2: 240891794-240891794 on Build GRCh38
rs766628931 Chr.2: 240891788-240891788 on Build GRCh38
rs760831770 Chr.2: 240891786-240891786 on Build GRCh38
rs772781785 Chr.2: 240891632-240891632 on Build GRCh38
rs78562453 Chr.2: 240891738-240891738 on Build GRCh38
rs527324672 Chr.2: 240891711-240891711 on Build GRCh38
rs201641583 Chr.2: 240891630-240891630 on Build GRCh38
rs753659085 Chr.2: 240891628-240891629 on Build GRCh38
rs375876316 Chr.2: 240891750-240891750 on Build GRCh38
rs752718874 Chr.2: 240891811-240891811 on Build GRCh38
rs750460148 Chr.2: 240891660-240891660 on Build GRCh38
rs769001091 Chr.2: 240891768-240891768 on Build GRCh38
rs746812384 Chr.2: 240891840-240891840 on Build GRCh38
rs774942147 Chr.2: 240891771-240891771 on Build GRCh38
rs201526003 Chr.2: 240891759-240891759 on Build GRCh38
rs774719912 Chr.2: 240891856-240891856 on Build GRCh38
rs753969212 Chr.2: 240891737-240891737 on Build GRCh38
rs765790974 Chr.2: 240891638-240891638 on Build GRCh38
rs201209605 Chr.2: 240891735-240891735 on Build GRCh38
rs777535104 Chr.2: 240891753-240891753 on Build GRCh38
rs758318525 Chr.2: 240891752-240891752 on Build GRCh38
rs757195248 Chr.2: 240891831-240891832 on Build GRCh38
rs551482681 Chr.2: 240891678-240891678 on Build GRCh38
rs372541574 Chr.2: 240891701-240891701 on Build GRCh38
rs756900249 Chr.2: 240891843-240891843 on Build GRCh38
rs779938737 Chr.2: 240891669-240891669 on Build GRCh38
rs202007238 Chr.2: 240891782-240891782 on Build GRCh38
rs749256148 Chr.2: 240891672-240891672 on Build GRCh38
rs781394354 Chr.2: 240891652-240891652 on Build GRCh38
rs765064683 Chr.2: 240891805-240891805 on Build GRCh38
rs761433613 Chr.2: 240891629-240891629 on Build GRCh38
rs762266508 Chr.2: 240891723-240891723 on Build GRCh38
rs201604905 Chr.2: 240891751-240891751 on Build GRCh38
rs371391472 Chr.2: 240891793-240891793 on Build GRCh38
rs768487561 Chr.2: 240891674-240891674 on Build GRCh38
rs555765305 Chr.2: 240891629-240891630 on Build GRCh38
rs368882820 Chr.2: 240891668-240891668 on Build GRCh38
rs776060776 Chr.2: 240891697-240891697 on Build GRCh38
rs771812388 Chr.2: 240891685-240891685 on Build GRCh38
rs750686303 Chr.2: 240891730-240891730 on Build GRCh38
rs766386211 Chr.2: 240891836-240891836 on Build GRCh38
rs1007195255 Chr.2: 240891710-240891710 on Build GRCh38
rs987835419 Chr.2: 240891696-240891696 on Build GRCh38
rs764671376 Chr.2: 240891702-240891702 on Build GRCh38
rs375967286 Chr.2: 240891728-240891728 on Build GRCh38
rs775942728 Chr.2: 240891760-240891760 on Build GRCh38
rs760239593 Chr.2: 240891693-240891693 on Build GRCh38
rs770604265 Chr.2: 240891695-240891695 on Build GRCh38
rs6737320 Chr.2: 240891847-240891847 on Build GRCh38
rs773023319 Chr.2: 240891689-240891689 on Build GRCh38

View Full Product Details