Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18450517-18450750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELL
Entrez Gene ID
8178
Gene Name
elongation factor for RNA polymerase II
Gene Aliases
C19orf17, ELL1, MEN, PPP1R68
UniGene
Hs.515260
Gene Chromosome Location
Chr.19: 18442663-18522129 on Build GRCh38
Transcripts
NM_006532.3 XM_017027335.1 XM_017027336.1 XM_017027337.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5765124 c.1241G>A p.G414E Chr.19: 18450701-18450701 on Build GRCh38
6653322 c.1400G>A p.R467Q Chr.19: 18450542-18450542 on Build GRCh38
166542 c.1276G>A p.G426S Chr.19: 18450666-18450666 on Build GRCh38
5755296 c.1213G>T p.G405C Chr.19: 18450729-18450729 on Build GRCh38
5733434 c.1331C>T p.S444L Chr.19: 18450611-18450611 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757187370 Chr.19: 18450702-18450702 on Build GRCh38
rs35318523 Chr.19: 18450566-18450566 on Build GRCh38
rs755783890 Chr.19: 18450671-18450671 on Build GRCh38
rs745557479 Chr.19: 18450611-18450611 on Build GRCh38
rs941620827 Chr.19: 18450660-18450660 on Build GRCh38
rs768028855 Chr.19: 18450633-18450633 on Build GRCh38
rs200447123 Chr.19: 18450666-18450666 on Build GRCh38
rs750718383 Chr.19: 18450580-18450580 on Build GRCh38
rs201725032 Chr.19: 18450672-18450672 on Build GRCh38
rs144216885 Chr.19: 18450621-18450621 on Build GRCh38
rs34078075 Chr.19: 18450610-18450610 on Build GRCh38
rs751440732 Chr.19: 18450696-18450696 on Build GRCh38
rs775807945 Chr.19: 18450544-18450544 on Build GRCh38
rs370344034 Chr.19: 18450550-18450550 on Build GRCh38
rs552804584 Chr.19: 18450679-18450679 on Build GRCh38
rs759014608 Chr.19: 18450543-18450543 on Build GRCh38
rs748811813 Chr.19: 18450674-18450674 on Build GRCh38
rs748692522 Chr.19: 18450678-18450678 on Build GRCh38
rs753948964 Chr.19: 18450649-18450649 on Build GRCh38
rs570976399 Chr.19: 18450677-18450677 on Build GRCh38
rs756997423 Chr.19: 18450665-18450665 on Build GRCh38
rs759620295 Chr.19: 18450694-18450694 on Build GRCh38
rs751965470 Chr.19: 18450560-18450560 on Build GRCh38
rs370308532 Chr.19: 18450650-18450650 on Build GRCh38
rs766612516 Chr.19: 18450684-18450684 on Build GRCh38
rs769123637 Chr.19: 18450616-18450616 on Build GRCh38
rs764296120 Chr.19: 18450559-18450559 on Build GRCh38
rs779727475 Chr.19: 18450710-18450710 on Build GRCh38
rs756110558 Chr.19: 18450590-18450590 on Build GRCh38
rs751528722 Chr.19: 18450664-18450664 on Build GRCh38
rs891827875 Chr.19: 18450592-18450592 on Build GRCh38
rs765321825 Chr.19: 18450695-18450695 on Build GRCh38
rs200244799 Chr.19: 18450642-18450642 on Build GRCh38
rs775049626 Chr.19: 18450630-18450630 on Build GRCh38
rs138181189 Chr.19: 18450608-18450608 on Build GRCh38
rs770568039 Chr.19: 18450542-18450542 on Build GRCh38
rs745441429 Chr.19: 18450667-18450667 on Build GRCh38
rs147064082 Chr.19: 18450600-18450600 on Build GRCh38
rs367661601 Chr.19: 18450609-18450609 on Build GRCh38
rs777756419 Chr.19: 18450605-18450605 on Build GRCh38
rs34868531 Chr.19: 18450663-18450663 on Build GRCh38
rs559281749 Chr.19: 18450603-18450603 on Build GRCh38
rs148295522 Chr.19: 18450622-18450622 on Build GRCh38
rs754492176 Chr.19: 18450715-18450715 on Build GRCh38
rs760965737 Chr.19: 18450641-18450641 on Build GRCh38
rs778584474 Chr.19: 18450721-18450721 on Build GRCh38
rs780206435 Chr.19: 18450598-18450598 on Build GRCh38
rs961060975 Chr.19: 18450626-18450626 on Build GRCh38
rs773522776 Chr.19: 18450636-18450636 on Build GRCh38
rs547360285 Chr.19: 18450703-18450703 on Build GRCh38
rs541916427 Chr.19: 18450631-18450631 on Build GRCh38
rs142159817 Chr.19: 18450564-18450564 on Build GRCh38
rs775066872 Chr.19: 18450619-18450619 on Build GRCh38
rs954933322 Chr.19: 18450657-18450657 on Build GRCh38
rs767832725 Chr.19: 18450572-18450572 on Build GRCh38

View Full Product Details