Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45396721-45396994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP27
Entrez Gene ID
201176
Gene Name
Rho GTPase activating protein 27
Gene Aliases
CAMGAP1, PP905, SH3D20, SH3P20
UniGene
Hs.205326
Gene Chromosome Location
Chr.17: 45393902-45432916 on Build GRCh38
Transcripts
NM_001282290.1 NM_199282.2 XM_005257128.3 XM_006721745.2 XM_006721747.2 XM_006721750.3 XM_011524468.1 XM_011524469.1 XM_011524470.2 XM_011524471.1 XM_011524472.2 XM_011524474.1 XM_017024311.1 XM_017024312.1 XM_017024313.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4662820 c.1943C>T p.P648L Chr.17: 45396924-45396924 on Build GRCh38
355769 c.915G>T p.A305A Chr.17: 45396929-45396929 on Build GRCh38
1183404 c.914C>T p.A305V Chr.17: 45396930-45396930 on Build GRCh38
4662818 c.1971C>A p.P657P Chr.17: 45396771-45396771 on Build GRCh38
6146909 c.1933G>A p.D645N Chr.17: 45396934-45396934 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765440494 Chr.17: 45396787-45396787 on Build GRCh38
rs749945885 Chr.17: 45396933-45396935 on Build GRCh38
rs186678479 Chr.17: 45396759-45396759 on Build GRCh38
rs758511206 Chr.17: 45396791-45396791 on Build GRCh38
rs754223102 Chr.17: 45396784-45396784 on Build GRCh38
rs766860936 Chr.17: 45396839-45396839 on Build GRCh38
rs751833468 Chr.17: 45396964-45396964 on Build GRCh38
rs148897544 Chr.17: 45396940-45396940 on Build GRCh38
rs192393004 Chr.17: 45396832-45396832 on Build GRCh38
rs200263300 Chr.17: 45396963-45396963 on Build GRCh38
rs767514153 Chr.17: 45396967-45396967 on Build GRCh38
rs755192707 Chr.17: 45396740-45396740 on Build GRCh38
rs777019676 Chr.17: 45396841-45396841 on Build GRCh38
rs776537416 Chr.17: 45396768-45396768 on Build GRCh38
rs1041898936 Chr.17: 45396864-45396864 on Build GRCh38
rs1040016522 Chr.17: 45396923-45396923 on Build GRCh38
rs376354313 Chr.17: 45396872-45396872 on Build GRCh38
rs777784651 Chr.17: 45396750-45396750 on Build GRCh38
rs772056533 Chr.17: 45396748-45396748 on Build GRCh38
rs375474591 Chr.17: 45396935-45396935 on Build GRCh38
rs747125169 Chr.17: 45396796-45396796 on Build GRCh38
rs779068048 Chr.17: 45396742-45396742 on Build GRCh38
rs773816294 Chr.17: 45396830-45396830 on Build GRCh38
rs202060202 Chr.17: 45396828-45396828 on Build GRCh38
rs993635880 Chr.17: 45396829-45396829 on Build GRCh38
rs779687119 Chr.17: 45396913-45396913 on Build GRCh38
rs149723446 Chr.17: 45396789-45396789 on Build GRCh38
rs763469092 Chr.17: 45396946-45396946 on Build GRCh38
rs778020696 Chr.17: 45396794-45396794 on Build GRCh38
rs755970723 Chr.17: 45396971-45396971 on Build GRCh38
rs745620936 Chr.17: 45396798-45396798 on Build GRCh38
rs373878834 Chr.17: 45396912-45396912 on Build GRCh38
rs778597676 Chr.17: 45396924-45396924 on Build GRCh38
rs773861287 Chr.17: 45396930-45396930 on Build GRCh38
rs764477136 Chr.17: 45396890-45396890 on Build GRCh38
rs7213200 Chr.17: 45396860-45396860 on Build GRCh38
rs199756859 Chr.17: 45396973-45396973 on Build GRCh38
rs748919047 Chr.17: 45396820-45396820 on Build GRCh38
rs1050348255 Chr.17: 45396969-45396969 on Build GRCh38
rs139027396 Chr.17: 45396976-45396976 on Build GRCh38
rs768564416 Chr.17: 45396909-45396909 on Build GRCh38
rs143628193 Chr.17: 45396965-45396965 on Build GRCh38
rs769483282 Chr.17: 45396812-45396812 on Build GRCh38
rs766907855 Chr.17: 45396783-45396783 on Build GRCh38
rs762606860 Chr.17: 45396941-45396941 on Build GRCh38
rs1030053633 Chr.17: 45396931-45396931 on Build GRCh38
rs900072522 Chr.17: 45396928-45396928 on Build GRCh38
rs780110494 Chr.17: 45396972-45396972 on Build GRCh38
rs745821061 Chr.17: 45396910-45396910 on Build GRCh38
rs757424661 Chr.17: 45396905-45396905 on Build GRCh38
rs770382952 Chr.17: 45396938-45396938 on Build GRCh38
rs764983476 Chr.17: 45396910-45396911 on Build GRCh38
rs755994713 Chr.17: 45396777-45396777 on Build GRCh38
rs201380561 Chr.17: 45396874-45396874 on Build GRCh38
rs140979770 Chr.17: 45396776-45396776 on Build GRCh38
rs755353135 Chr.17: 45396785-45396785 on Build GRCh38
rs574227892 Chr.17: 45396889-45396889 on Build GRCh38
rs773665081 Chr.17: 45396780-45396780 on Build GRCh38
rs757257402 Chr.17: 45396797-45396797 on Build GRCh38
rs889110305 Chr.17: 45396774-45396774 on Build GRCh38
rs769476480 Chr.17: 45396771-45396771 on Build GRCh38
rs748243517 Chr.17: 45396743-45396743 on Build GRCh38
rs571177508 Chr.17: 45396818-45396818 on Build GRCh38
rs372242163 Chr.17: 45396927-45396927 on Build GRCh38
rs763629512 Chr.17: 45396778-45396778 on Build GRCh38
rs761351117 Chr.17: 45396954-45396954 on Build GRCh38
rs368827618 Chr.17: 45396770-45396770 on Build GRCh38
rs145583506 Chr.17: 45396929-45396929 on Build GRCh38
rs751675861 Chr.17: 45396900-45396900 on Build GRCh38
rs749117763 Chr.17: 45396915-45396915 on Build GRCh38
rs373045289 Chr.17: 45396782-45396782 on Build GRCh38
rs746973778 Chr.17: 45396756-45396756 on Build GRCh38
rs761353583 Chr.17: 45396768-45396769 on Build GRCh38

View Full Product Details