Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 2736144-2736387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTN4
Entrez Gene ID
152330
Gene Name
contactin 4
Gene Aliases
AXCAM, BIG-2
UniGene
Hs.298705
Gene Chromosome Location
Chr.3: 2098771-3059080 on Build GRCh38
Transcripts
NM_001206955.1 NM_175607.2 XM_006713004.3 XM_011533425.2 XM_011533426.2 XM_011533427.2 XM_011533428.2 XM_011533429.2 XM_011533430.2 XM_011533431.2 XM_017005782.1 XM_017005783.1 XM_017005784.1 XM_017005785.1 XM_017005786.1 XM_017005787.1 XM_017005788.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6631791 c.130delA p.V46fs*1 Chr.3: 2736289-2736289 on Build GRCh38
4673315 c.77C>A p.P26Q Chr.3: 2736236-2736236 on Build GRCh38
7199698 c.131A>G p.K44R Chr.3: 2736290-2736290 on Build GRCh38
1043474 c.129G>T p.E43D Chr.3: 2736288-2736288 on Build GRCh38
1043473 c.91G>T p.E31* Chr.3: 2736250-2736250 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748853919 Chr.3: 2736257-2736257 on Build GRCh38
rs750631515 Chr.3: 2736351-2736351 on Build GRCh38
rs956532658 Chr.3: 2736294-2736294 on Build GRCh38
rs751434014 Chr.3: 2736176-2736176 on Build GRCh38
rs753979733 Chr.3: 2736303-2736303 on Build GRCh38
rs777082764 Chr.3: 2736334-2736334 on Build GRCh38
rs774231861 Chr.3: 2736260-2736260 on Build GRCh38
rs759480723 Chr.3: 2736186-2736186 on Build GRCh38
rs567978105 Chr.3: 2736262-2736262 on Build GRCh38
rs867810602 Chr.3: 2736221-2736221 on Build GRCh38
rs752738447 Chr.3: 2736189-2736189 on Build GRCh38
rs758736193 Chr.3: 2736325-2736325 on Build GRCh38
rs34786854 Chr.3: 2736195-2736196 on Build GRCh38
rs780448288 Chr.3: 2736326-2736326 on Build GRCh38
rs772802775 Chr.3: 2736245-2736245 on Build GRCh38
rs755541221 Chr.3: 2736332-2736332 on Build GRCh38
rs777946573 Chr.3: 2736215-2736215 on Build GRCh38
rs748844430 Chr.3: 2736338-2736338 on Build GRCh38
rs746232520 Chr.3: 2736234-2736234 on Build GRCh38
rs146175469 Chr.3: 2736236-2736236 on Build GRCh38
rs375343574 Chr.3: 2736237-2736237 on Build GRCh38
rs779206846 Chr.3: 2736226-2736226 on Build GRCh38
rs767486743 Chr.3: 2736267-2736267 on Build GRCh38
rs767576704 Chr.3: 2736187-2736187 on Build GRCh38
rs536718929 Chr.3: 2736274-2736274 on Build GRCh38
rs556639444 Chr.3: 2736305-2736305 on Build GRCh38
rs747428832 Chr.3: 2736331-2736331 on Build GRCh38
rs921295660 Chr.3: 2736315-2736315 on Build GRCh38
rs774141536 Chr.3: 2736354-2736354 on Build GRCh38
rs775536329 Chr.3: 2736271-2736271 on Build GRCh38
rs765454741 Chr.3: 2736313-2736313 on Build GRCh38
rs528403275 Chr.3: 2736222-2736222 on Build GRCh38
rs756143199 Chr.3: 2736190-2736190 on Build GRCh38
rs769168400 Chr.3: 2736243-2736243 on Build GRCh38
rs770712694 Chr.3: 2736258-2736258 on Build GRCh38
rs753984004 Chr.3: 2736217-2736217 on Build GRCh38
rs750714691 Chr.3: 2736323-2736323 on Build GRCh38
rs968013770 Chr.3: 2736310-2736310 on Build GRCh38
rs988004711 Chr.3: 2736298-2736298 on Build GRCh38
rs764028050 Chr.3: 2736281-2736281 on Build GRCh38

View Full Product Details