Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18541814-18542065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP8
Entrez Gene ID
23770
Gene Name
FK506 binding protein 8
Gene Aliases
FKBP38, FKBPr38
UniGene
Hs.173464
Gene Chromosome Location
Chr.19: 18531752-18544077 on Build GRCh38
Transcripts
NM_001308373.1 NM_012181.4 XM_011527863.1 XM_011527864.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1630775 c.135A>G p.E45E Chr.19: 18541836-18541836 on Build GRCh38
4684789 c.48C>T p.A16A Chr.19: 18541923-18541923 on Build GRCh38
993123 c.46G>A p.A16T Chr.19: 18541925-18541925 on Build GRCh38
3701432 c.126A>C p.E42D Chr.19: 18541845-18541845 on Build GRCh38
5431749 c.105A>G p.E35E Chr.19: 18541953-18541953 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764311952 Chr.19: 18541884-18541884 on Build GRCh38
rs142246175 Chr.19: 18541902-18541902 on Build GRCh38
rs760871342 Chr.19: 18542020-18542020 on Build GRCh38
rs758835267 Chr.19: 18541962-18541962 on Build GRCh38
rs10854166 Chr.19: 18542034-18542034 on Build GRCh38
rs763320647 Chr.19: 18541981-18541981 on Build GRCh38
rs933677129 Chr.19: 18541871-18541871 on Build GRCh38
rs933275119 Chr.19: 18541899-18541899 on Build GRCh38
rs778744089 Chr.19: 18541846-18541848 on Build GRCh38
rs1035933416 Chr.19: 18542036-18542036 on Build GRCh38
rs768417071 Chr.19: 18541837-18541848 on Build GRCh38
rs375921391 Chr.19: 18541907-18541907 on Build GRCh38
rs772769071 Chr.19: 18541850-18541850 on Build GRCh38
rs373061427 Chr.19: 18541908-18541908 on Build GRCh38
rs760545001 Chr.19: 18542029-18542029 on Build GRCh38
rs775982766 Chr.19: 18541978-18541978 on Build GRCh38
rs750485111 Chr.19: 18542014-18542014 on Build GRCh38
rs371498399 Chr.19: 18541922-18541922 on Build GRCh38
rs922813849 Chr.19: 18541842-18541842 on Build GRCh38
rs372625663 Chr.19: 18541923-18541923 on Build GRCh38
rs747615730 Chr.19: 18541855-18541866 on Build GRCh38
rs376029668 Chr.19: 18541912-18541912 on Build GRCh38
rs543806511 Chr.19: 18541963-18541963 on Build GRCh38
rs532658624 Chr.19: 18542009-18542009 on Build GRCh38
rs764322218 Chr.19: 18542004-18542004 on Build GRCh38
rs760166092 Chr.19: 18541911-18541911 on Build GRCh38
rs776172610 Chr.19: 18541867-18541867 on Build GRCh38
rs11541428 Chr.19: 18541925-18541925 on Build GRCh38
rs771709504 Chr.19: 18541841-18541841 on Build GRCh38
rs760028266 Chr.19: 18541870-18541870 on Build GRCh38
rs371561631 Chr.19: 18541863-18541863 on Build GRCh38
rs770683600 Chr.19: 18541935-18541935 on Build GRCh38
rs763260727 Chr.19: 18541877-18541877 on Build GRCh38
rs773138745 Chr.19: 18541998-18541998 on Build GRCh38
rs185670325 Chr.19: 18541851-18541851 on Build GRCh38
rs746623839 Chr.19: 18541926-18541926 on Build GRCh38
rs765196636 Chr.19: 18542033-18542033 on Build GRCh38
rs368293221 Chr.19: 18542005-18542005 on Build GRCh38
rs573142562 Chr.19: 18541872-18541872 on Build GRCh38
rs1011017418 Chr.19: 18542038-18542038 on Build GRCh38
rs563584235 Chr.19: 18541999-18541999 on Build GRCh38
rs201298401 Chr.19: 18541944-18541944 on Build GRCh38
rs948537674 Chr.19: 18541941-18541941 on Build GRCh38
rs771517020 Chr.19: 18541994-18541995 on Build GRCh38
rs767904202 Chr.19: 18542012-18542012 on Build GRCh38
rs539205072 Chr.19: 18541896-18541896 on Build GRCh38

View Full Product Details