Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1978982-1979219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1G2
Entrez Gene ID
1455
Gene Name
casein kinase 1 gamma 2
Gene Aliases
CK1g2
UniGene
Hs.651905
Gene Chromosome Location
Chr.19: 1941149-1981338 on Build GRCh38
Transcripts
NM_001319.6 XM_005259498.1 XM_005259499.3 XM_005259500.1 XM_005259501.2 XM_017026297.1 XM_017026298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4076029 c.614C>T p.P205L Chr.19: 1979025-1979025 on Build GRCh38
4076028 c.611T>C p.I204T Chr.19: 1979022-1979022 on Build GRCh38
4650461 c.692G>A p.R231H Chr.19: 1979172-1979172 on Build GRCh38
3531160 c.636G>A p.L212L Chr.19: 1979047-1979047 on Build GRCh38
3531158 c.589G>A p.D197N Chr.19: 1979000-1979000 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566024183 Chr.19: 1979157-1979157 on Build GRCh38
rs768784883 Chr.19: 1979108-1979108 on Build GRCh38
rs760408468 Chr.19: 1979100-1979101 on Build GRCh38
rs139042120 Chr.19: 1979017-1979017 on Build GRCh38
rs774654686 Chr.19: 1979119-1979119 on Build GRCh38
rs35546534 Chr.19: 1979141-1979142 on Build GRCh38
rs763741937 Chr.19: 1979072-1979072 on Build GRCh38
rs991097765 Chr.19: 1979125-1979125 on Build GRCh38
rs770330378 Chr.19: 1979113-1979113 on Build GRCh38
rs758389451 Chr.19: 1979104-1979104 on Build GRCh38
rs779803565 Chr.19: 1979106-1979106 on Build GRCh38
rs55754218 Chr.19: 1979060-1979060 on Build GRCh38
rs757105761 Chr.19: 1979084-1979084 on Build GRCh38
rs773593161 Chr.19: 1979114-1979114 on Build GRCh38
rs771483023 Chr.19: 1979118-1979118 on Build GRCh38
rs748367551 Chr.19: 1979112-1979112 on Build GRCh38
rs751538407 Chr.19: 1979147-1979147 on Build GRCh38
rs56340103 Chr.19: 1979030-1979030 on Build GRCh38
rs781397404 Chr.19: 1979158-1979158 on Build GRCh38
rs1023378186 Chr.19: 1979135-1979135 on Build GRCh38
rs370181224 Chr.19: 1979059-1979059 on Build GRCh38
rs56038081 Chr.19: 1979079-1979079 on Build GRCh38
rs112874571 Chr.19: 1979105-1979105 on Build GRCh38
rs763584907 Chr.19: 1979120-1979120 on Build GRCh38
rs750208966 Chr.19: 1979101-1979102 on Build GRCh38
rs375946759 Chr.19: 1979032-1979032 on Build GRCh38
rs781628669 Chr.19: 1979109-1979109 on Build GRCh38
rs770275176 Chr.19: 1979015-1979015 on Build GRCh38
rs56231501 Chr.19: 1979045-1979045 on Build GRCh38
rs763179754 Chr.19: 1979115-1979115 on Build GRCh38
rs539881059 Chr.19: 1979179-1979179 on Build GRCh38
rs761426026 Chr.19: 1979121-1979121 on Build GRCh38
rs763492411 Chr.19: 1979025-1979025 on Build GRCh38
rs748418954 Chr.19: 1979164-1979164 on Build GRCh38
rs999413228 Chr.19: 1979107-1979107 on Build GRCh38
rs56356031 Chr.19: 1979050-1979050 on Build GRCh38
rs951919122 Chr.19: 1979155-1979155 on Build GRCh38
rs752275442 Chr.19: 1979056-1979056 on Build GRCh38
rs758067532 Chr.19: 1979129-1979129 on Build GRCh38
rs55841667 Chr.19: 1979053-1979053 on Build GRCh38
rs750369892 Chr.19: 1979102-1979102 on Build GRCh38
rs528244206 Chr.19: 1979124-1979124 on Build GRCh38
rs373703759 Chr.19: 1979134-1979134 on Build GRCh38
rs547896883 Chr.19: 1979146-1979146 on Build GRCh38
rs56264133 Chr.19: 1979028-1979028 on Build GRCh38
rs56108438 Chr.19: 1979027-1979027 on Build GRCh38
rs55818316 Chr.19: 1979033-1979033 on Build GRCh38
rs750252577 Chr.19: 1979128-1979128 on Build GRCh38
rs937901679 Chr.19: 1979110-1979111 on Build GRCh38

View Full Product Details