Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97922285-97922544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMO
Entrez Gene ID
51531
Gene Name
tRNA methyltransferase O
Gene Aliases
C9orf156, HSPC219, NAP1
UniGene
Hs.9196
Gene Chromosome Location
Chr.9: 97876321-97922570 on Build GRCh38
Transcripts
NM_016481.4 XM_005252047.3 XM_005252048.4 XM_005252049.4 XM_005252050.4 XM_011518776.2 XM_011518777.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
455018 c.19T>C p.S7P Chr.9: 97922475-97922475 on Build GRCh38
3763479 c.13T>C p.S5P Chr.9: 97922475-97922475 on Build GRCh38
3763480 c.16T>C p.S6P Chr.9: 97922475-97922475 on Build GRCh38
6616843 c.36A>G p.T12T Chr.9: 97922458-97922458 on Build GRCh38
3763477 c.51T>G p.V17V Chr.9: 97922437-97922437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977368237 Chr.9: 97922330-97922330 on Build GRCh38
rs756586569 Chr.9: 97922450-97922450 on Build GRCh38
rs766164192 Chr.9: 97922479-97922479 on Build GRCh38
rs746189244 Chr.9: 97922405-97922405 on Build GRCh38
rs759453936 Chr.9: 97922485-97922485 on Build GRCh38
rs775767150 Chr.9: 97922468-97922468 on Build GRCh38
rs746950691 Chr.9: 97922383-97922383 on Build GRCh38
rs199804891 Chr.9: 97922496-97922496 on Build GRCh38
rs746111324 Chr.9: 97922462-97922462 on Build GRCh38
rs1003730279 Chr.9: 97922454-97922454 on Build GRCh38
rs879545148 Chr.9: 97922514-97922514 on Build GRCh38
rs959335416 Chr.9: 97922309-97922309 on Build GRCh38
rs867758231 Chr.9: 97922456-97922456 on Build GRCh38
rs969031874 Chr.9: 97922324-97922324 on Build GRCh38
rs766106334 Chr.9: 97922446-97922446 on Build GRCh38
rs377466224 Chr.9: 97922376-97922376 on Build GRCh38
rs1022715159 Chr.9: 97922477-97922477 on Build GRCh38
rs150014670 Chr.9: 97922469-97922469 on Build GRCh38
rs970705746 Chr.9: 97922377-97922377 on Build GRCh38
rs749477329 Chr.9: 97922395-97922395 on Build GRCh38
rs932433735 Chr.9: 97922382-97922382 on Build GRCh38
rs372240346 Chr.9: 97922501-97922501 on Build GRCh38
rs1031940161 Chr.9: 97922307-97922307 on Build GRCh38
rs374859311 Chr.9: 97922513-97922513 on Build GRCh38
rs3183928 Chr.9: 97922437-97922437 on Build GRCh38
rs781749940 Chr.9: 97922502-97922502 on Build GRCh38
rs368353816 Chr.9: 97922489-97922489 on Build GRCh38
rs778886130 Chr.9: 97922491-97922491 on Build GRCh38
rs769321252 Chr.9: 97922512-97922512 on Build GRCh38
rs772433249 Chr.9: 97922465-97922465 on Build GRCh38
rs964372825 Chr.9: 97922445-97922445 on Build GRCh38
rs749696407 Chr.9: 97922516-97922517 on Build GRCh38
rs952737057 Chr.9: 97922430-97922430 on Build GRCh38
rs187366296 Chr.9: 97922407-97922407 on Build GRCh38
rs143058977 Chr.9: 97922303-97922303 on Build GRCh38
rs771207689 Chr.9: 97922398-97922398 on Build GRCh38
rs369570790 Chr.9: 97922499-97922499 on Build GRCh38
rs752586451 Chr.9: 97922522-97922522 on Build GRCh38
rs894926078 Chr.9: 97922490-97922490 on Build GRCh38
rs747848285 Chr.9: 97922306-97922306 on Build GRCh38
rs764924410 Chr.9: 97922433-97922433 on Build GRCh38
rs894832511 Chr.9: 97922315-97922315 on Build GRCh38
rs1011940603 Chr.9: 97922320-97922320 on Build GRCh38
rs769537109 Chr.9: 97922331-97922331 on Build GRCh38
rs760579392 Chr.9: 97922523-97922523 on Build GRCh38
rs780410911 Chr.9: 97922469-97922470 on Build GRCh38
rs777876778 Chr.9: 97922391-97922391 on Build GRCh38
rs767335042 Chr.9: 97922520-97922520 on Build GRCh38
rs761542305 Chr.9: 97922432-97922432 on Build GRCh38
rs915273506 Chr.9: 97922321-97922321 on Build GRCh38
rs776323798 Chr.9: 97922429-97922429 on Build GRCh38
rs139234146 Chr.9: 97922449-97922449 on Build GRCh38
rs376128350 Chr.9: 97922453-97922453 on Build GRCh38
rs761108572 Chr.9: 97922427-97922427 on Build GRCh38
rs369881999 Chr.9: 97922455-97922455 on Build GRCh38
rs111419479 Chr.9: 97922313-97922313 on Build GRCh38
rs924406983 Chr.9: 97922334-97922334 on Build GRCh38
rs756133526 Chr.9: 97922385-97922385 on Build GRCh38
rs532293548 Chr.9: 97922367-97922367 on Build GRCh38
rs3183927 Chr.9: 97922475-97922475 on Build GRCh38
rs758588929 Chr.9: 97922494-97922494 on Build GRCh38
rs372086299 Chr.9: 97922458-97922458 on Build GRCh38
rs777289698 Chr.9: 97922473-97922473 on Build GRCh38
rs758172189 Chr.9: 97922442-97922442 on Build GRCh38
rs3824496 Chr.9: 97922363-97922363 on Build GRCh38
rs200242824 Chr.9: 97922464-97922464 on Build GRCh38
rs943900048 Chr.9: 97922344-97922344 on Build GRCh38
rs569967200 Chr.9: 97922342-97922342 on Build GRCh38
rs565535676 Chr.9: 97922518-97922518 on Build GRCh38
rs1000579982 Chr.9: 97922305-97922305 on Build GRCh38
rs764684064 Chr.9: 97922474-97922474 on Build GRCh38
rs370149200 Chr.9: 97922381-97922381 on Build GRCh38
rs779127924 Chr.9: 97922460-97922460 on Build GRCh38
rs762557863 Chr.9: 97922478-97922478 on Build GRCh38
rs906663591 Chr.9: 97922450-97922450 on Build GRCh38
rs952539856 Chr.9: 97922519-97922519 on Build GRCh38
rs370852825 Chr.9: 97922486-97922486 on Build GRCh38
rs574170108 Chr.9: 97922471-97922471 on Build GRCh38
rs774568709 Chr.9: 97922402-97922402 on Build GRCh38
rs748686651 Chr.9: 97922505-97922505 on Build GRCh38
rs1012319293 Chr.9: 97922493-97922493 on Build GRCh38
rs776030045 Chr.9: 97922410-97922410 on Build GRCh38
rs751488458 Chr.9: 97922451-97922451 on Build GRCh38
rs148772479 Chr.9: 97922482-97922482 on Build GRCh38
rs200865938 Chr.9: 97922487-97922487 on Build GRCh38

View Full Product Details