Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 98891478-98891711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGF1R
Entrez Gene ID
3480
Gene Name
insulin like growth factor 1 receptor
Gene Aliases
CD221, IGFIR, IGFR, JTK13
UniGene
Hs.643120
Gene Chromosome Location
Chr.15: 98648539-98964530 on Build GRCh38
Transcripts
NM_000875.4 NM_001291858.1 XM_011521516.2 XM_017022136.1 XM_017022137.1 XM_017022138.1 XM_017022139.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7160752 c.814G>C p.E272Q Chr.15: 98891498-98891498 on Build GRCh38
6261930 c.880G>T p.E294* Chr.15: 98891564-98891564 on Build GRCh38
5520229 c.943G>A p.G315S Chr.15: 98891627-98891627 on Build GRCh38
3957088 c.810G>C p.R270S Chr.15: 98891494-98891494 on Build GRCh38
5020636 c.864C>T p.A288A Chr.15: 98891548-98891548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754913548 Chr.15: 98891515-98891515 on Build GRCh38
rs780190444 Chr.15: 98891654-98891654 on Build GRCh38
rs756249894 Chr.15: 98891647-98891647 on Build GRCh38
rs915169594 Chr.15: 98891623-98891623 on Build GRCh38
rs750031695 Chr.15: 98891497-98891497 on Build GRCh38
rs886051559 Chr.15: 98891579-98891579 on Build GRCh38
rs45460595 Chr.15: 98891503-98891503 on Build GRCh38
rs1030757824 Chr.15: 98891579-98891579 on Build GRCh38
rs56303650 Chr.15: 98891569-98891569 on Build GRCh38
rs755253910 Chr.15: 98891657-98891657 on Build GRCh38
rs770848753 Chr.15: 98891527-98891527 on Build GRCh38
rs752567548 Chr.15: 98891628-98891628 on Build GRCh38
rs371115707 Chr.15: 98891670-98891670 on Build GRCh38
rs766118129 Chr.15: 98891620-98891620 on Build GRCh38
rs2229764 Chr.15: 98891587-98891587 on Build GRCh38
rs767871468 Chr.15: 98891684-98891684 on Build GRCh38
rs34524380 Chr.15: 98891655-98891655 on Build GRCh38
rs747107423 Chr.15: 98891524-98891524 on Build GRCh38
rs778765650 Chr.15: 98891520-98891520 on Build GRCh38
rs769196604 Chr.15: 98891545-98891545 on Build GRCh38
rs774791538 Chr.15: 98891546-98891546 on Build GRCh38
rs375731039 Chr.15: 98891645-98891645 on Build GRCh38
rs775798637 Chr.15: 98891539-98891539 on Build GRCh38
rs369004098 Chr.15: 98891646-98891646 on Build GRCh38
rs201304073 Chr.15: 98891557-98891557 on Build GRCh38
rs766174659 Chr.15: 98891563-98891563 on Build GRCh38
rs144675711 Chr.15: 98891534-98891534 on Build GRCh38
rs778482528 Chr.15: 98891671-98891671 on Build GRCh38
rs927703464 Chr.15: 98891633-98891633 on Build GRCh38
rs776224615 Chr.15: 98891681-98891681 on Build GRCh38
rs201529410 Chr.15: 98891564-98891564 on Build GRCh38
rs45540536 Chr.15: 98891669-98891669 on Build GRCh38
rs2229764 Chr.15: 98891587-98891587 on Build GRCh38
rs779108826 Chr.15: 98891665-98891665 on Build GRCh38
rs570009181 Chr.15: 98891622-98891622 on Build GRCh38
rs758297620 Chr.15: 98891629-98891629 on Build GRCh38
rs748557435 Chr.15: 98891599-98891599 on Build GRCh38
rs201956526 Chr.15: 98891627-98891627 on Build GRCh38
rs34430460 Chr.15: 98891548-98891548 on Build GRCh38
rs772820424 Chr.15: 98891613-98891613 on Build GRCh38
rs774794966 Chr.15: 98891588-98891588 on Build GRCh38
rs566063029 Chr.15: 98891581-98891581 on Build GRCh38
rs755846821 Chr.15: 98891500-98891500 on Build GRCh38
rs780195678 Chr.15: 98891586-98891586 on Build GRCh38
rs554696305 Chr.15: 98891614-98891614 on Build GRCh38
rs200954736 Chr.15: 98891554-98891554 on Build GRCh38
rs746040080 Chr.15: 98891531-98891531 on Build GRCh38
rs56385625 Chr.15: 98891683-98891683 on Build GRCh38
rs745984927 Chr.15: 98891582-98891582 on Build GRCh38
rs35171849 Chr.15: 98891530-98891530 on Build GRCh38
rs547142975 Chr.15: 98891573-98891573 on Build GRCh38
rs374358935 Chr.15: 98891507-98891507 on Build GRCh38
rs760366319 Chr.15: 98891558-98891558 on Build GRCh38
rs748565822 Chr.15: 98891666-98891666 on Build GRCh38
rs770508100 Chr.15: 98891677-98891677 on Build GRCh38
rs55954954 Chr.15: 98891626-98891626 on Build GRCh38
rs772738892 Chr.15: 98891611-98891611 on Build GRCh38
rs756302851 Chr.15: 98891585-98891585 on Build GRCh38

View Full Product Details