Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 27495675-27495924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMCO2
Entrez Gene ID
341346
Gene Name
single-pass membrane protein with coiled-coil domains 2
Gene Aliases
C12orf70
UniGene
Hs.282121
Gene Chromosome Location
Chr.12: 27446482-27506911 on Build GRCh38
Transcripts
NM_001145010.1 XM_011520634.2 XM_011520635.2 XM_011520636.2 XM_017019254.1 XM_017019255.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4518817 c.668_669CC>TT p.P223L Chr.12: 27495840-27495841 on Build GRCh38
4819323 c.740C>T p.S247F Chr.12: 27495762-27495762 on Build GRCh38
6287091 c.688G>T p.A230S Chr.12: 27495710-27495710 on Build GRCh38
4819325 c.590C>T p.S197F Chr.12: 27495762-27495762 on Build GRCh38
6838234 c.566C>T p.A189V Chr.12: 27495738-27495738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964486961 Chr.12: 27495741-27495741 on Build GRCh38
rs758477943 Chr.12: 27495753-27495753 on Build GRCh38
rs897049322 Chr.12: 27495806-27495806 on Build GRCh38
rs910417901 Chr.12: 27495730-27495730 on Build GRCh38
rs909456030 Chr.12: 27495827-27495827 on Build GRCh38
rs771726659 Chr.12: 27495852-27495852 on Build GRCh38
rs1013707085 Chr.12: 27495854-27495854 on Build GRCh38
rs1031519549 Chr.12: 27495720-27495720 on Build GRCh38
rs1048252441 Chr.12: 27495838-27495838 on Build GRCh38
rs994130264 Chr.12: 27495816-27495816 on Build GRCh38
rs1046439482 Chr.12: 27495876-27495876 on Build GRCh38
rs779685023 Chr.12: 27495784-27495784 on Build GRCh38
rs778993662 Chr.12: 27495812-27495812 on Build GRCh38
rs749160521 Chr.12: 27495788-27495788 on Build GRCh38
rs306646 Chr.12: 27495853-27495853 on Build GRCh38
rs547964063 Chr.12: 27495727-27495727 on Build GRCh38
rs751493883 Chr.12: 27495769-27495769 on Build GRCh38
rs759937888 Chr.12: 27495709-27495709 on Build GRCh38
rs3751225 Chr.12: 27495891-27495891 on Build GRCh38
rs566187249 Chr.12: 27495772-27495772 on Build GRCh38
rs546963781 Chr.12: 27495810-27495810 on Build GRCh38
rs988931407 Chr.12: 27495710-27495710 on Build GRCh38
rs957545495 Chr.12: 27495722-27495722 on Build GRCh38
rs544816724 Chr.12: 27495897-27495897 on Build GRCh38
rs35515888 Chr.12: 27495846-27495847 on Build GRCh38
rs754957022 Chr.12: 27495790-27495790 on Build GRCh38
rs752770562 Chr.12: 27495743-27495743 on Build GRCh38
rs1047444114 Chr.12: 27495785-27495785 on Build GRCh38
rs3751223 Chr.12: 27495777-27495777 on Build GRCh38
rs1049810871 Chr.12: 27495773-27495773 on Build GRCh38
rs61997182 Chr.12: 27495844-27495844 on Build GRCh38
rs185962940 Chr.12: 27495778-27495778 on Build GRCh38

View Full Product Details