Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150267485-150267754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APH1A
Entrez Gene ID
51107
Gene Name
aph-1 homolog A, gamma-secretase subunit
Gene Aliases
6530402N02Rik, APH-1, APH-1A, CGI-78
UniGene
Hs.108408
Gene Chromosome Location
Chr.1: 150265399-150282291 on Build GRCh38
Transcripts
NM_001077628.2 NM_001243771.1 NM_001243772.1 NM_016022.3 NR_045033.1 NR_045034.1 NR_045035.1 XM_017001417.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4662445 c.332C>T p.P111L Chr.1: 150267742-150267742 on Build GRCh38
4021947 c.322G>A p.G108R Chr.1: 150267752-150267752 on Build GRCh38
4662444 c.332C>T p.P111L Chr.1: 150267742-150267742 on Build GRCh38
4021946 c.322G>A p.G108R Chr.1: 150267752-150267752 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs974409962 Chr.1: 150267630-150267630 on Build GRCh38
rs16835635 Chr.1: 150267637-150267637 on Build GRCh38
rs782815079 Chr.1: 150267610-150267610 on Build GRCh38
rs958161951 Chr.1: 150267622-150267622 on Build GRCh38
rs782242981 Chr.1: 150267678-150267678 on Build GRCh38
rs781950430 Chr.1: 150267701-150267701 on Build GRCh38
rs112212494 Chr.1: 150267715-150267715 on Build GRCh38
rs587710793 Chr.1: 150267646-150267646 on Build GRCh38
rs782401232 Chr.1: 150267674-150267674 on Build GRCh38
rs782170334 Chr.1: 150267729-150267729 on Build GRCh38
rs782610145 Chr.1: 150267691-150267691 on Build GRCh38
rs925239465 Chr.1: 150267536-150267536 on Build GRCh38
rs202225606 Chr.1: 150267704-150267704 on Build GRCh38
rs587684750 Chr.1: 150267677-150267677 on Build GRCh38
rs940216052 Chr.1: 150267714-150267714 on Build GRCh38
rs782021342 Chr.1: 150267718-150267718 on Build GRCh38
rs373046159 Chr.1: 150267699-150267699 on Build GRCh38
rs369637410 Chr.1: 150267509-150267509 on Build GRCh38
rs587629635 Chr.1: 150267671-150267671 on Build GRCh38

View Full Product Details